Skip to Content
Merck
All Photos(1)

Key Documents

GE90100422

HiTrap® IgM Purification

pkg of 1 × 5 mL, Cytiva, 90-1004-22

Sign Into View Organizational & Contract Pricing

Select a Size

5 ML
$1,730.00

$1,730.00


Check Cart for Availability


Select a Size

Change View
5 ML
$1,730.00

About This Item

UNSPSC Code:
41106500
NACRES:
NB.21

$1,730.00


Check Cart for Availability

packaging

pkg of 1 × 5 mL

manufacturer/tradename

90-1004-22
Cytiva

Related Categories

Legal Information

HiTrap is a registered trademark of Cytiva

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service