Skip to Content
Merck
All Photos(1)

Key Documents

GE90-1003-97

HiTrap® 5X1ML IGG Sepharose 6FF

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106500
NACRES:
NA.56
Product GE90-1003-97 is not currently sold in your country. Contact Technical Service

manufacturer/tradename

Cytiva

bed size

7 mm × NA

bed volume

1 mL

column I.D.

(7 mm)

matrix

highly cross-linked 6% agarose

median particle size

90 μm

cleaning

3-10

working range

3-10

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.
Store at 4 to 30 °C (20% Ethanol)

Legal Information

HiTrap is a registered trademark of Cytiva

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Storage Class Code

3 - Flammable liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service