Skip to Content
Merck
All Photos(1)

Key Documents

GE90100123

Cytiva (TM) Blue Sepharose High Performance

Sign Into View Organizational & Contract Pricing

Select a Size

50 ML
$2,930.00

$2,930.00


Check Cart for Availability


Select a Size

Change View
50 ML
$2,930.00

About This Item

UNSPSC Code:
23151817
NACRES:
SB.53

$2,930.00


Check Cart for Availability

packaging

pack of 50 mL

manufacturer/tradename

Cytiva 90100123

Legal Information

Sepharose is a trademark of Cytiva

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

Flash Point(F)

55.4 °F

Flash Point(C)

13 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service