Key Documents
GE80648546
GE Ettan DALTsix gel caster
volume 1000 mL, pkg of 1-6 gels
About This Item
UNSPSC Code:
41105300
NACRES:
NB.22
packaging
pack of 1 ea
pkg of 1-6 gels
manufacturer/tradename
Cytiva 80-6485-46
Cytiva Article No. 80-6485-46
volume
1000 mL
General description
DALTsix Gel Caster holds up to six 1.0-mm or 1.5-mm cassettes with separator sheets for casting homogeneous or gradient gels. If fewer gels are to be cast, the unused volume is filled with blank cassette inserts and filler sheets.
Legal Information
Ettan is a trademark of Cytiva
Choose from one of the most recent versions:
Certificates of Analysis (COA)