GE80-6484-32
Mini Dialysis Kit, 8 kDa
Cytiva 80-6484-32, pack of 2 mL
About This Item
Recommended Products
feature
wetted part: no
packaging
pack of 2 mL
manufacturer/tradename
Cytiva 80-6484-32
storage temp.
2-8°C
Related Categories
General description
Application
The dialysis tubes in the kit each have a cap that incorporates a small disk of dialysis membrane. Samples are easily pipetted into and removed from the conical bottom of the tube. The capped tube is inserted into a float and then, with the cap and membrane down, placed in a stirred beaker containing the desired dialysis solution.
Following dialysis, the tube is centrifuged briefly. This brings the entire contents of the tube back to the conical bottom for maximum recovery. The dialyzed sample can be stored in the same tube by simply replacing the dialysis cap with a standard cap.
Features and Benefits
- Designed for efficient dialysis of small sample volumes.
- Uses disposable dialysis tubes with dialysis membrane incorporated into caps.
- Conical tube bottom maximizes sample recovery
Storage and Stability
Analysis Note
Storage Class Code
12 - Non Combustible Liquids
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Protocols
Mini Dialysis Kit from Cytiva minimizes sample handling and loss, reducing streaking on 2-D gels from contaminants.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service