Skip to Content
Merck
All Photos(2)

Key Documents

GE29343057

Sera-Mag Select

Cytiva 29343057, pack of 1 × 450 mL

Sign Into View Organizational & Contract Pricing

Select a Size

450 ML
$5,600.00

$5,600.00


Check Cart for Availability


Select a Size

Change View
450 ML
$5,600.00

About This Item

UNSPSC Code:
41105507
NACRES:
NB.13

$5,600.00


Check Cart for Availability

packaging

pack of 1 × 450 mL

manufacturer/tradename

Cytiva 29343057

avg. part. size

1 μm

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

Sera-Mag Select PCR clean-up and size selection reagent combines the exceptional binding characteristics of Sera-Mag Carboxyl Speedbeads with an optimized binding solution in a ready-to-use formulation. The DNA size range isolated from the process can be tailored to suit the user′s end requirements by adjusting the amount of reagent that is mixed with a fixed volume of sample.

  • Use a single reagent for PCR clean-up and size selection.
  • Easily control what size DNA is retained or eliminated by adding various ratios of Sera-Mag Select to your sample. This enables NGS library and PCR fragment clean-up, as well as dual size selection of libraries.
  • Uses the same protocols as existing reagents so can be directly integrated into existing workflows with minimal disruption.
  • Sera-Mag Select delivers tight distribution in size selection for high yield recovery of specific fragment sizes.
  • Suitable for manual, high throughput and automated workflows.
  • Sera-Mag Select should be stored in the refrigerator between 2°C to 8°C. Do not freeze. Store upright and keep bottle tightly sealed.

Legal Information

Sera-Mag is a trademark of Cytiva

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service