Skip to Content
Merck
All Photos(1)

Key Documents

GE29-0486-65

HiTrap Protein L

Cytiva 29-0486-65, pack of 1 mL

Synonym(s):

Protein L chromatography

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
23151816
NACRES:
NA.56
Product GE29-0486-65 is not currently sold in your country. Contact Technical Service

ligand

recombinant protein L (E. coli), mammalian free

packaging

pack of 1 mL

manufacturer/tradename

Cytiva 29-0486-65

availability

not available in North America

storage condition

(20% Ehtanol)

parameter

5 bar

column I.D.

7 mm

matrix

rigid, highly cross-linked agarose

avg. part. size

85 μm (d₅₀v is the median particle size of the cumulative volume distribution)

working range

2-10(pH interval where the medium can be operated without significant change in function.)

suitability

suitable for bioprocess medium

storage temp.

2-8°C

Related Categories

General description

HiTrap™ Protein L columns are prepacked with Capto™ L affinity chromatography resin for purification of antibody fragments containing the kappa light chain. This resin combines a rigid, high-flow agarose matrix with the immunoglobulin-binding recombinant protein L ligand, which has a strong affinity to the variable region of antibody kappa light chains.

Application

Affinity chromatoghraphy; capture of antibodies and antibody fragments such as Fab fragments, scFv, and Dabs.

Features and Benefits

  • Suitable for the capture of a wide range of antibody fragments such as antigen-binding fragments (Fabs), domain antibodies (dAbs), and single-chain variable fragments (scFvs).
  • Enables a platform approach to antibody fragment purification.
  • High binding capacity, low ligand leakage, and selectivity for a broad range of antibody fragments reduces process time and amount of resin.
  • Convenient for process development, as well as small-scale purification.
  • HiTrap™ format enables easy connection to a syringe, peristaltic pump, or chromatography system such as an ÄKTA™ system.

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.

Legal Information

HisTrap is a trademark of Cytiva

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Storage Class Code

3 - Flammable liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Philipp A Kronenberg et al.
Frontiers in cellular and infection microbiology, 13, 1162530-1162530 (2023-04-04)
Alveolar (AE) and cystic echinococcosis (CE) are severe parasitic zoonoses caused by the larval stages of Echinococcus multilocularis and E. granulosus sensu lato, respectively. A panel of 7 monoclonal antibodies (mAbs) was selected against major diagnostic epitopes of both species.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service