Skip to Content
Merck
All Photos(1)

Key Documents

GE28-9523-61

GST Buffer Kit

Cytiva 28-9523-61, pack of 1 kit

Sign Into View Organizational & Contract Pricing

Select a Size

1 PKG
$821.00

$821.00


Check Cart for Availability


Select a Size

Change View
1 PKG
$821.00

About This Item

UNSPSC Code:
41106501
NACRES:
NA.56

$821.00


Check Cart for Availability

sterility

non-sterile

packaging

pack of 1 kit

manufacturer/tradename

Cytiva 28-9523-61

shipped in

wet ice

storage temp.

2-8°C

Related Categories

General description

GST Buffer Kit contains carefully prepared buffer concentrates for binding, washing, and elution of GST-tagged proteins according to recommended protocols. The kit contains 10× PBS, reduced glutathione, dilution buffer, and an instruction booklet. Sufficient reagents are supplied to purify up to 20 mg of GST-tagged protein

GST Buffer Kit eliminates time-consuming buffer preparation and therefore allows fast, reproducible, and convenient purification work. GST Buffer Kit can be used in combination with a number of column types: GST SpinTrap, GST MultiTrap 96-well plates, GST GraviTrap, GSTrap FF, GSTrap 4B and GSTrap HP.

Features and Benefits

  • Premade stock solutions of buffers optimized for GST-tagged protein purification.

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.

Analysis Note

To view the Certificate of Analysis for this product, please visit www.cytiva.com.

Storage Class Code

12 - Non Combustible Liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Articles

This page shows how to purify GST-tagged proteins using GST SpinTrap™ from Cytiva.

This page shows troubleshooting methods for purification of GST-tagged proteins using Cytiva products.

This page describes characteristics of Glutathione Sepharose and HiTrap® Benzamidine FF (high sub) media and columns from Cytiva.

Protocols

This page provides information about different pull-down assays for the further isolation of multiprotein complexes to identify their components with products from Cytiva.

Related Content

Investigate in vitro protein-protein interactions with pull-down assays, utilizing affinity, GST pull-down, TAP, and co-immunoprecipitation methods.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service