GE28-4065-51
dNTP Set (100 mM each A,C,G,T)
Cytiva 28-4065-51, pack of 4 × 25 μmol
Select a Size
$343.00
Select a Size
About This Item
$343.00
Recommended Products
form
liquid
packaging
pack of 4 × 25 μmol
manufacturer/tradename
Cytiva 28-4065-51
concentration
100 mM
color
Colorless
shipped in
dry ice
storage temp.
−20°C
Related Categories
General description
Features and Benefits
- Free from DNase, RNase, and nicking enzyme activity
- Greater than 99% triphosphate purity (by HPLC) for optimum performance and consistency
- Buffer free and ready-to-use solutions in multiple formats
- Functionally tested for long PCR* and sequencing
- Custom formulation, testing, packaging, and labeling are available
Storage and Stability
Analysis Note
Legal Information
Storage Class Code
12 - Non Combustible Liquids
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service