Key Documents
GE28-4064-16
Sigma-Aldrich
Tricorn 10/150 Column
Select a Size
1 EACH
$1,280.00
$1,280.00
Check Cart for Availability
Request a Bulk Order
Select a Size
Change View
1 EACH
$1,280.00
About This Item
UNSPSC Code:
41121800
NACRES:
SB.52
$1,280.00
Check Cart for Availability
Request a Bulk Order
technique(s)
LPLC: suitable
General description
Tricorn 5 and 10 Coarse Filter Kit is used for large volume/repeated loading of clarified sample to reduce the risk of clogging in combination with capture media. Both filter and coarse filter kits include top and bottom adapters, bottom filters, and EPDM O-rings, five of each.
Tricorn Packing Equipment includes the Packing Equipment 5/50 (5-mm connector and 50-mm glass tube) or 10/100 (10-mm connector and 100-mm glass tube), EPDM O-rings and bottom unit, cap, and stop plug.
Features and Benefits
Broad range of column dimensions suitable for many different purification needs.
Glass tubes coated with plastic film prevent harm if the column is dropped or operated outside its specification.
Locking ring prevents accidental compression of media bed.
Separate packing connector allows simple column packing for a broad range of media.
The modern integrated, snap-on design results in a column that is easy to assembly, operate, and maintain.
Tricorn empty columns are designed for high performance separations on small and medium scale. Pressure stability up to 100 bar and durable column material ensure reliable and reproducible operation as well as compatibility with the majority of the commercial lab media.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Visit the Document Library
Articles
The different stages of working with SOURCE™ (Cytiva) for high-resolution separation are discussed, from purification to polishing and scaling up
This page covers practical problems that may lead to a non-ideal IEX separation and their solutions.
This page covers detailed aspects of each step in an IEX separation to improve resolution and overall performance.
Protocols
Here we discuss purification options when working with Sepharose® High Performance and we present some examples of media screening, capture and purification.
This page covers the use of Sepharose High Performance media for purification of proteins, peptides or oligonucleotides, when to use them, and with which systems.
This page discusses how to do a purification with SOURCE™ media from Cytiva.
Column Packing and Preparation for Size Exclusion Chromatography
See All