Skip to Content
Merck
All Photos(1)

Key Documents

GE17549111

HiTrap® MabSelect SuRe pcc

Cytiva 175491111, pkg of 5 × 1 mL

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106500
NACRES:
NB.21
Product GE17549111 is not currently sold in your country. Contact Technical Service

packaging

pkg of 5 × 1 mL

manufacturer/tradename

Cytiva 175491111

bed H × I.D. × volume

(25 mm) (7 mm) (1 mL)

shipped in

ambient

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

HiTrap MabSelect SuRe pcc chromatography columns are prepacked with MabSelect SuRe pcc protein A resin for screening and small-scale mAb purification in continuous bioprocessing using periodic counter-current chromatography (PCC).

  • Convenient prepacked column for fast, reproducible, and easy small-scale purifications.
  • High binding capacity, ~ 60 mg human IgG/mL resin at 2.4 min residence time
  • High productivity through capture of large mass of monoclonal antibodies in a short period of time.
  • Highly concentrated elution pools for operating flexibility and small unit operations.
  • Allows cost-effective cleaning with 0.1–0.5 M NaOH over hundreds of purification cycles.
  • High protein ligand stability reduces ligand leakage.

HiTrap MabSelect SuRe pcc packed columns (1 mL and 5 mL) are suited for small scale antibody purifications, as well as optimization of methods and parameters, such as sample load and protein binding conditions. These protein A columns provide fast, reproducible, and easy separations in a convenient format. The columns are optimally utilized when they are connected to liquid chromatography systems such as the ÄKTA systems.

MabSelect SuRe pcc affinity resin offers exceptional capacity at short residence time, making it well-suited for applications requiring fast mass transfer such as monoclonal antibody capture in a continuous process.

The alkali-stabilized, protein A-derived ligand gives the resin greater stability than conventional protein A-based resins under the alkaline conditions used in cleaning-in-place (CIP) protocols. Consequently, the possibility of cleaning with cost-effective reagents such as sodium hydroxide (NaOH) will improve process economy.

In addition, the use of NaOH for cleaning efficiently inactivates bacteria, mold, and yeast, with improved product quality as a result.

Features and Benefits

  • Convenient prepacked column for Fast, reproducible, and easy small-scale purifications
  • High binding capacity, ~ 60 mg human IgG/mL resin at 2.4 min residence time
  • High productivity through capture of large mass of monoclonal antibodies in a short period of time
  • Highly concentrated elution pools for operating flexibility and small unit operations
  • Allows cost-effective cleaning with 0.1–0.5 M NaOH over hundreds of purification cycles
  • High protein ligand stability reduces ligand leakage

Legal Information

HiTrap is a registered trademark of Cytiva
MabSelect SuRe is a trademark of Cytiva

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service