Skip to Content
Merck
All Photos(1)

Key Documents

GE17-5484-10

HiScreen Capto Phenyl ImpRes

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
23151817
NACRES:
NA.56
Product GE17-5484-10 is not currently sold in your country. Contact Technical Service

bed size

7.7 mm × 100 mm

bed volume

4.7 mL

column I.D.

7.7 mm

General description

HiScreen Capto Phenyl ImpRes columns are prepacked with Capto Phenyl ImpRes hydrophobic interaction chromatography medium, and are part of the process development platform available from Cytiva. The columns are an excellent choice for method optimization and parameter screening.

  • 10 cm bed height of HiScreen columns is designed to allow method optimization and parameter screening
  • Easily connected in series to achieve 20 cm bed height
  • Small bed volume gives fast results and minimal sample/buffer consumption
  • Reproducible results, scalable to BioProcess columns packed with the same chromatography media using the same linear fluid velocity

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.

Legal Information

HiScreen is a trademark of Cytiva

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Storage Class Code

3 - Flammable liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service