Skip to Content
Merck
All Photos(1)

Key Documents

GE17-1271-01

Cytopore 2

Cytiva, dry powder

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41161502
NACRES:
NC.01
Product GE17-1271-01 is not currently sold in your country. Contact Technical Service

feature

dry powder

manufacturer/tradename

17-1271-01
Cytiva

Legal Information

Cytopore is a trademark of Cytiva

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service