Skip to Content
Merck
All Photos(1)

Key Documents

GE17-0575-01

Chelating Sepharose Fast Flow

Cytiva 17-0575-01, pack of 50 mL

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106500
NACRES:
NA.56

ligand

iminodiacetic acid

packaging

pack of 50 mL

manufacturer/tradename

Cytiva 17-0575-01

matrix

6% cross-linked agarose

particle size

45-165 μm

average diameter

90 μm

cleaning in place

2-14

working range

3-13

suitability

suitable for bioprocess medium

General description

Chelating Sepharose Fast Flow is a BioProcess IMAC medium, designed for purifying native and recombinant proteins and peptides that have an affinity for metal ions.

Chelating Sepharose Fast Flow is composed of cross-linked 6% agarose beads modified with iminodiacetic immobilized to the base matrix by stable ether linkages and sufficiently long spacer arms.

Chelating Sepharose Fast Flow is supplied free of metal ions, allowing the user to charge it with the most appropriate metal ion for purification of a target protein.

Chelating Sepharose Fast Flow is available in a range of different bulk pack sizes for easy scale-up and process development. As member of the BioProcess media range, Chelating Sepharose Fast Flow meets industrial demands with security of supply and comprehensive technical and regulatory support

Features and Benefits

  • Uncharged IMAC resin allowing possibility to charge with your metal ion of choice for optimized selectivity
  • Convenient purification of native and recombinant proteins
  • As BioProcess medium it fulfills industrial demands for security of supply, robust performance and regulatory support.
  • BioProcess medium supported for industrial applications and well-established in approved processes

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.
Store at 4 to 30 °C (20% Ethanol)

Analysis Note

To view the Certificate of Analysis for this product, please visit www.cytiva.com.

Legal Information

Sepharose is a trademark of Cytiva

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Storage Class Code

3 - Flammable liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

How to separate proteins and peptides with affinity for metal ions by immobilized metal chelate affinity chromatography.

This page shows how to purify or remove proteins and peptides with exposed amino acids with Chelating Sepharose High Performance, Chelating Sepharose Fast Flow, Capto Chelating from Cytiva.

This page shows how to convert between flow velocity and volumetric flow rate in affinity chromatography of antibodies.

Protocols

Guide for separating poly(His) fusion proteins using affinity chromatography tags in various purification products.

Sample Preparation for Affinity Chromatography in Specific Groups of Biomolecules

This page shows how to convert between linear flow and volumetric flow rates in affinity chromatography.

How to perform buffer exchange and desalting with Sephadex G-25, HiTrap Desalting columns, or ÄKTAprime plus.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service