GE10600121
Amersham™ Hybond® P Western blotting membrane sandwich, PVDF
pore size 0.45 μm, sheet W × L 80 mm × 90 mm , preassembled with 2 x 3MM chr filter papers, pkg of 10 ea
Select a Size
$189.00
Select a Size
About This Item
$189.00
Recommended Products
packaging
pkg of 10 ea
manufacturer/tradename
Cytiva 10600121
sheet W × L
80 mm × 90 mm , preassembled with 2 x 3MM chr filter papers
pore size
0.45 μm pore size
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Features and Benefits
- Precut Amersham Hybond P 0.45 µm PVDF blotting membrane.
Storage and Stability
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Articles
This page describes troubleshooting strategies in the detection of GST-tagged proteins.
Protocols
This page describes how to monitor expression and purification of GST-tagged proteins by Western blot analysis using Cytiva products.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service