Skip to Content
Merck
All Photos(2)

Key Documents

GBL622202

Grace Bio-Labs CoverWell perfusion chambers

1 chamber, port diam. 1.5 mm, chamber W × L × thickness 19 mm × 32 mm × 2.5 mm

Synonym(s):

leak proof enclosure for slides, microscope stick on multiwell chamber, reusable press to seal multiwell chamber, water tight multiple well chamber

Sign Into View Organizational & Contract Pricing

Select a Size

20 EA
$261.00

$261.00


Check Cart for Availability


Select a Size

Change View
20 EA
$261.00

About This Item

UNSPSC Code:
41122600
NACRES:
NB.22

$261.00


Check Cart for Availability

material

clear polycarbonate backing
red silicon wells

feature

wells 1

packaging

pack of 20 ea

manufacturer/tradename

Grace Bio-Labs 622202

chamber W × L × thickness

19 mm × 32 mm × 2.5 mm

port diam.

1.5 mm

wells

1

Looking for similar products? Visit Product Comparison Guide

General description

CoverWell perfusion chambers press-to-seal covers form water-tight, multiwell cell incubation or cytochemistry chambers when pressed to coverslips or microscope slides. Reagents can be quickly added and removed through dual access ports without disturbing or cross-contaminating specimens. Chambers are available in five chamber configurations allowing up to eight different specimens or analytical tests on one coverglass.

CoverWell perfusion chambers form removable, water-tight multiwell enclosures by pressing to coverglass or microscope slides. Chambers are made of medical-grade silicone bonded to clear, UV-transparent plastic, and are removable for permanent mounting. The thin polycarbonate cover displays excellent transparency. The perfusion chambers are ideal for live cell imaging on upright or inverted microscopes at high magnification.

Legal Information

CoverWell is a trademark of Grace Bio-Labs, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service