Skip to Content
Merck
All Photos(2)

Key Documents

GBL611202

Grace Bio-Labs HybriWell-FL sealing system, Fluor-"friendly" adhesive chamber

chamber volume 140-165 μL, L × W × thickness 22 mm × 60 mm × 0.12 mm, port diam. 1.5 mm

Synonym(s):

chamber slides, chambered coverglass, cover slides, hybridization chambers, incubation chambers

Sign Into View Organizational & Contract Pricing

Select a Size

100 EA
$649.00

$649.00


Check Cart for Availability


Select a Size

Change View
100 EA
$649.00

About This Item

UNSPSC Code:
41122600
NACRES:
NB.22

$649.00


Check Cart for Availability

material

colorless (black adhesive)
polycarbonate

feature

wells 1

packaging

pack of 100 ea

manufacturer/tradename

Grace Bio-Labs 611202

L × W × thickness

22 mm × 60 mm × 0.12 mm

chamber volume

140-165 μL

port diam.

1.5 mm

wells

1

Looking for similar products? Visit Product Comparison Guide

General description

Grace Bio-Labs HybriWell sealing system seals securely to a microscope slide surface in seconds to accommodate water bath hybridizations, such as protein or nucleic acid hybridization assays. The HybriWell fluorescent (HybriWell-FL) hybridization system has been designed specifically for sensitive DNA microarray assays involving Cy®5 or Alexa Fluor 647 direct-labeled DNA probes.

Features:
  • Encloses a single or multiple specimens in a small reagent volume and eliminate evaporation.
  • Access ports in the chamber surface allow for the addition or removal of reactants.
  • Ports are easily sealed using adhesive seal tabs provided with each order.
  • Sealed chambers are watertight and ideally suited for waterbath incubations.
  • RNase free, hydrophobic surfaces will not trap or bind probes.
  • Disposable chamber removes cleanly and easily even after heating.
  • Each package includes illustrated instructions for use, applicator and 200 adhesive port seals.

Legal Information

ALEXA FLUOR is a trademark of Life Technologies
Cy is a registered trademark of Cytiva
HybriWell is a trademark of Grace Bio-Labs, Inc.
HybriWell-FL is a trademark of Grace Bio-Labs, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service