Key Documents
GBL505196
Grace Bio-Labs ONCYTE® NOVA nitrocellulose film slides
96 square pads, 6 mm diam.glass substrate
Synonym(s):
nitrocellulose slides, protein microarrays
Select a Size
1 EA
$425.00
$425.00
Check Cart for Availability
Select a Size
Change View
1 EA
$425.00
About This Item
UNSPSC Code:
41122600
NACRES:
NB.15
$425.00
Check Cart for Availability
material
nitrocellulose (on glass)
packaging
pack of 1 ea
manufacturer/tradename
Grace Bio-Labs 505196
external size
74 mm × 110 mm
binding type
medium binding surface
General description
Grace Bio-Labs ONCYTE® NOVA nitrocellulose coated substrates are designed for SBS compliant multiwell plates and glass substrates for high throughput automated processing of microarrays.
Legal Information
NOVA is a trademark of Grace Bio-Labs, Inc.
Oncyte is a registered trademark of Grace Bio-Labs, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)