Skip to Content
Merck
All Photos(2)

Key Documents

GBL246880

Grace Bio-Labs ProPlate® microarray system tray set

size 16 wells, round, with stainless steel spring clips

Synonym(s):

incubation chambers, microarray hybridization chambers, slide chambers

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$314.00

$314.00


Check Cart for Availability


Select a Size

Change View
1 EA
$314.00

About This Item

UNSPSC Code:
41122600
NACRES:
NB.15

$314.00


Check Cart for Availability

material

black silicone (Acrylic tray)
round wells

sterility

non-sterile

feature

wells 16

packaging

pack of 1 ea

manufacturer/tradename

Grace Bio-Labs 246880

size

16 wells

well volume

350 μL

color

none

Looking for similar products? Visit Product Comparison Guide

General description

ProPlate re-usable, multi-well chambers form removable, leak-proof wells on virtually any slide surface without the use of adhesive.
Designs feature ANSI-SLAS compliant well spacing to facilitate multichannel pipetting.
Multiple slides insert easily into frames having a standard microtiter plate footprint for automated processing.

Legal Information

ProPlate is a registered trademark of Grace Bio-Labs, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service