Key Documents
GBL204878
GRACE BIO-LABS SEALSTRIPS AND SEALSTRIP APPLICATOR, FOR PROPLATE® MICROARRAY SYSTEMS
for ProPlate® microarray systems, pack of 1
Synonym(s):
microarray system applicator, seal strip applicator
Select a Size
1 EA
$27.30
$27.30
Check Cart for Availability
Select a Size
Change View
1 EA
$27.30
About This Item
UNSPSC Code:
41122600
NACRES:
NB.17
$27.30
Check Cart for Availability
feature
for ProPlate® microarray systems
packaging
pack of 1
manufacturer/tradename
Grace Bio-Labs 204878
General description
This plastic applicator helps to apply seal strips to the top surface of the chambers. Seal strips are ideal to prevent evaporation, they remove cleanly and easily without leaving a residue.
Legal Information
ProPlate is a registered trademark of Grace Bio-Labs, Inc.
SealStrips is a trademark of Grace Bio-Labs, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)