Key Documents
GBL204837
Grace Bio-Labs ProPlate® clips for microarray systems
stainless steel numbered, pack of 2 ea
Synonym(s):
microarray clips, stainless steel spring clips
Select a Size
2 EA
$27.30
$27.30
Check Cart for Availability
Select a Size
Change View
2 EA
$27.30
About This Item
UNSPSC Code:
41121800
NACRES:
NB.17
$27.30
Check Cart for Availability
feature
stainless steel numbered
packaging
pack of 2 ea
manufacturer/tradename
Grace Bio-Labs 204837
General description
Grace Bio-Labs ProPlate® multi-array chambers (microarray systems) are designed to be used with ONCYTE® nitrocellulose-coated slides. These multi-array chambers integrate microscope slide based microarray technology with high-throughput (HTS applications) microtiter plate processing. Up to 64 individual arrays can be assayed per slide.
Three or four slides fit into a tray (depending on clip choice). The resulting multi-array chamber tray set produces a modular plate with a standard microtiter plate footprint and well spacing allowing high-throughput robotic processing of up to 256 arrays.
Features:
  • Up to 64 leak-proof wells on any slide surface without the use of adhesive
  • Microtiter plate well spacing facilitates use with multichannel pipettes
  • 7 x 7 mm square wells are ideal for high content arrays 5-350 ul well volumes accommodate small reagent and large wash volumes
ProPlate® stainless steel Spring Clips are easily applied and removed and are preferred by customers using glass slides fabricated by means of a laser and which may have small micro-fractures from processing or chipped edges. Stainless steel Spring Clips are numbered and autoclavable.
Legal Information
Oncyte is a registered trademark of Grace Bio-Labs, Inc.
ProPlate is a registered trademark of Grace Bio-Labs, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)