Skip to Content
Merck
All Photos(2)

Key Documents

G9920

Sigma-Aldrich

Anti-GABAB Receptor 2 antibody produced in rabbit

affinity isolated antibody, lyophilized powder

Synonym(s):

Anti-γ-Aminobutyric acid receptor type B subunit 2, Anti-GABABR2

Sign Into View Organizational & Contract Pricing

Select a Size

50 μL
$472.00
200 μL
$981.00

$472.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
50 μL
$472.00
200 μL
$981.00

About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

$472.00


Check Cart for Availability

Request a Bulk Order

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

lyophilized powder

species reactivity

rat

technique(s)

western blot: 1:200 using rat brain membranes

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

Related Categories

General description

The GABAB receptor (GABABR) is a heteromeric G-protein-coupled receptor. It has two subunits-GABAB1 (γ-aminobutyric acid type B receptor subunit 1) and GABAB2 (γ-aminobutyric acid type B receptor subunit 2). In the hippocampus, GABAB2 is present in dendrites and located in the extra-synaptic membrane of spines and dendritic shafts. This gene is located on rat chromosome 5q.

Immunogen

peptide CKDPIEDINSPEHIQRRLSL corresponding to residues 875-894 of human GABABR2.

Application

Anti-GABAB Receptor 2 antibody produced in rabbit is suitable for western blotting at a working dilution of 1:200 using rat brain membranes.
Anti-GABAB Receptor 2 antibody has been used in western blotting.
Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Western Blotting (1 paper)

Biochem/physiol Actions

The inhibitory neurotransmitter GABA signals through two distinct types of pre- and postsynaptic receptors, GABAA and GABAB. Both GABA receptors can regulate depression of synaptic transmission and be involved in the inhibition controlling neuronal excitability. The GABA transmitter interacts with three types of receptors: the ionotropic receptors GABAA and GABAC and the metabotropic GABAB receptor (GABABR). GABABR is a G-protein coupled receptor that plays a key role in slow synaptic inhibition in the brain and spinal cord. GABABR is a heteromer with two subunits, GABABR1 and GABABR2. The amino acids within the second intracellular domain of GABA(B2) are essential for the coupling of GABA(B) receptor heterodimers to their downstream effector systems. The N-termini on both the GB1 and GB2 receptors are involved in the normal coupling of GABAB receptors to their physiological effectors, G(i) and G-protein-activated potassium channels (GIRKs).

Target description

The GABABR2 was shown to be essential for trafficking and G-protein binding, while the GABAB R1 subunit was demonstrated to be important for agonist and antagonist binding. GABABR2 is highly expressed in brain especially at the cerebral cortex, thalamus, hippocampus, and spinal cord and to a lesser extent in testis.

Physical form

Lyophilized from phosphate buffered saline, pH 7.4, containing 1% BSA and 0.05% sodium azide.

Reconstitution

Reconstitute the lyophilized vial with 50 μL or 200 μL deionized water, depending on package size. Further dilutions should be made using a carrier protein such as BSA (1-3%).

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Margeta-Mitrovic et al.
Proceedings of the National Academy of Sciences of the United States of America, 98(25), 14643-14648 (2001-11-29)
gamma-aminobutyric acid type B (GABA(B)) receptors, G protein-coupled receptors (GPCRs) for GABA, are obligate heterodimers of two homologous subunits, GB1 and GB2. Typical for family C GPCRs, the N termini of both GB1 and GB2 contain a domain with homology
M J Robbins et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 21(20), 8043-8052 (2001-10-06)
GABA(B) receptors are unique among G-protein-coupled receptors (GPCRs) in their requirement for heterodimerization between two homologous subunits, GABA(B1) and GABA(B2), for functional expression. Whereas GABA(B1) is capable of binding receptor agonists and antagonists, the role of each GABA(B) subunit in
Mathias Boller et al.
Journal of neurophysiology, 89(4), 2035-2045 (2003-04-11)
In mammals, GABA(C) receptors seem to be specifically expressed in the retina and the subcortical visual system, with highest extraretinal expression levels in the superior colliculus (SC). Although its presence in the superficial SC has been demonstrated physiologically, a direct
M Margeta-Mitrovic et al.
Proceedings of the National Academy of Sciences of the United States of America, 98(25), 14649-14654 (2001-11-29)
Many G protein-coupled receptors (GPCRs) have recently been shown to dimerize, and it was suggested that dimerization may be a prerequisite for G protein coupling. gamma-aminobutyric acid type B (GABA(B)) receptors (GPCRs for GABA, a major inhibitory neurotransmitter in the
Yinghui Guo et al.
Experimental and therapeutic medicine, 9(4), 1448-1454 (2015-03-18)
The aim of the present study was to investigate the genes associated with 'anger-in' (tendency to suppress anger) and 'anger-out' (tendency to express anger through verbal or physical means) emotions in humans. Wistar rats were divided into five groups (n=10/group)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service