Skip to Content
Merck
All Photos(1)

Key Documents

G9894

Sigma-Aldrich

γ-Globulins from mouse

≥90% (agarose gel electrophoresis)

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$766.00
50 MG
$2,650.00

$766.00


Estimated to ship on17 April 2025

Diagnostics Manufacturers should use alternative grade product: SRE0011

Request a Bulk Order

Select a Size

Change View
10 MG
$766.00
50 MG
$2,650.00

About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.61

$766.00


Estimated to ship on17 April 2025

Diagnostics Manufacturers should use alternative grade product: SRE0011

Request a Bulk Order

biological source

mouse

Quality Level

Assay

≥90% (agarose gel electrophoresis)

form

powder

technique(s)

ELISA: suitable

impurities

≤4% NaCl

storage temp.

−20°C

Gene Information

mouse ... ggld(14591)

General description

γ-Globulin molecules is made up of four polypeptide chains, of which two have molecular weights of about 50,000 and two of about 20,000. These chains are cross-linked by disulfide bonds.[1]

Application

γ-Globulins from mouse were used to test the development and validation of ELISA for Herceptin detection in human serum.[2]

Preparation Note

Prepared from Cohn Fraction II, III

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Carola W N Damen et al.
Analytical biochemistry, 391(2), 114-120 (2009-05-26)
Trastuzumab, a humanized monoclonal antibody, is used for the treatment of breast cancer patients who overexpress the HER2 receptor. To optimize therapy, pharmacokinetic studies are necessary. The aim of this study was to develop an enzyme-linked immunosorbent assay (ELISA) for
[Postoperative thrombosis prevention using low-dose heparin].
A E Schaer et al.
Archiv fur Gynakologie, 219(1-4), 545-545 (1975-11-18)
Farhan Aslam et al.
Journal of medicinal food, 20(5), 448-457 (2017-03-24)
White sesame seed oil (WSSO) has been used in cooking and food preparations for centuries. It has many purported health benefits and may be a promising nutraceutical. The primary purpose of this study was to examine the effects of WSSO

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service