Skip to Content
Merck
All Photos(1)

Key Documents

G9665

Sigma-Aldrich

Anti-Glutamate Receptor 1A, Metabotropic (mGluR1A) antibody produced in rabbit

affinity isolated antibody, buffered aqueous glycerol solution

Sign Into View Organizational & Contract Pricing

Select a Size

0.1 ML
$606.00

$606.00


Estimated to ship on28 May 2025



Select a Size

Change View
0.1 ML
$606.00

About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

$606.00


Estimated to ship on28 May 2025


biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous glycerol solution

species reactivity

mouse, feline, rat

technique(s)

immunohistochemistry (frozen sections): 1:500
western blot: 1:1,000

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

mouse ... Grm1(14816)
rat ... Grm1(24414)

General description

The metabotropic glutamate receptors consist of eight subtypes (mGluR1-8) divided into three groups (I-III). Glutamate is the main excitatory neurotransmitter in the brain. It acts on ligand-gated receptor channels, termed NMDA, AMPA and kainate receptors, involved in the fast excitatory synaptic transmission. Glutamate also regulates ion channels and enzymes producing second messengers via specific receptors coupled to G-proteins, called metabotropic glutamate receptors.

Specificity

This antibody does not react with other splice variants or mGluR5.

Immunogen

synthetic peptide corresponding to the sequence of amino acids 1116-1130 of rat metabotropic glutamate receptor 1A (mGluR1a).

Application

Anti-Glutamate Receptor 1A, Metabotropic (mGluR1A) antibody produced in rabbit is suitable for use as a primary antibody at a working dilution of 1:1500 using extracts from mouse brain It is suitable for immunohistochemistry at a working dilution of 1:500 and for immunoblotting at a working dilution of 1:1000.

Biochem/physiol Actions

Glutamate Receptor 1 is a G-protein coupled receptor for glutamate. The role of the metabotropic receptors in pain and pathology is being studied for therapeutic potential. Glutamate receptors are important mediators of excitatory amino acid neurotransmission. The α isomer of this receptor is a disulfide-linked homodimer and its activity is mediated by a G-protein-coupled phosphatidylinositol-calcium second messenger system. Mutations in this gene causes autosomal-recessive congenital cerebellar ataxia.

Physical form

10 mM HEPES (pH 7.5), 150 mM NaCl, 100 μg/mL BSA and 50% glycerol

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sonja Janmaat et al.
The Journal of comparative neurology, 512(4), 467-477 (2008-11-26)
In vivo, homozygous staggerer (Rora(sg/sg)) Purkinje cells (PCs) remain in an early stage of development with rudimentary spineless dendrites, associated with a lack of parallel fiber (PF) input and the persistence of multiple climbing fibers (CFs). In this immunocytochemical study
Huixian Wu et al.
Science (New York, N.Y.), 344(6179), 58-64 (2014-03-08)
The excitatory neurotransmitter glutamate induces modulatory actions via the metabotropic glutamate receptors (mGlus), which are class C G protein-coupled receptors (GPCRs). We determined the structure of the human mGlu1 receptor seven-transmembrane (7TM) domain bound to a negative allosteric modulator, FITM
Tatjana Surdin et al.
iScience, 26(1), 105828-105828 (2023-01-13)
Neuronal plasticity underlying cerebellar learning behavior is strongly associated with type 1 metabotropic glutamate receptor (mGluR1) signaling. Activation of mGluR1 leads to activation of the Gq/11 pathway, which is involved in inducing synaptic plasticity at the parallel fiber-Purkinje cell synapse
Kunie Sakurai et al.
Developmental neurobiology, 69(12), 811-824 (2009-08-13)
The neural cell recognition molecule NB-3, also referred to as contactin-6, is expressed prominently in the developing nervous system after birth and its deficiency has been shown to cause impairment in motor coordination. Here, we investigated the contribution of NB-3
Velina Guergueltcheva et al.
American journal of human genetics, 91(3), 553-564 (2012-08-21)
Autosomal-recessive congenital cerebellar ataxia was identified in Roma patients originating from a small subisolate with a known strong founder effect. Patients presented with global developmental delay, moderate to severe stance and gait ataxia, dysarthria, mild dysdiadochokinesia, dysmetria and tremors, intellectual

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service