Skip to Content
Merck
All Photos(1)

Key Documents

G9398

Sigma-Aldrich

Asialoganglioside-GM2 from bovine brain

~95%, lyophilized powder

Synonym(s):

GA2 Ganglioside, Gangliotriosyl ceramide

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.77

Assay

~95%

Quality Level

form

lyophilized powder

storage temp.

−20°C

General description

Gangliosides are major constituents of neuronal cell membranes and endoplasmic reticulum; contain a sialated polysaccharide chain linked to ceramide through a β-glycosidic linkage; for classification of gangliosides see Svennerholm, L., et al. (eds.), Structure and Function of Gangliosides, New York, Plenum, 1980.

Biochem/physiol Actions

Degraded form of Asialoganglioside-GM1 ganglioside.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Christian H Schweppe et al.
Journal of lipid research, 51(8), 2282-2294 (2010-05-07)
Shiga toxin (Stx)-producing Escherichia coli are the leading cause of hemorrhagic colitis and life-threatening extraintestinal complications in humans. Stx1 and Stx2 are transferred by yet to be delineated mechanisms from the intestine to the circulation where they injure microvascular endothelial
Aufaugh Emam et al.
Microbiology (Reading, England), 152(Pt 9), 2789-2799 (2006-09-02)
The glycosphingolipids (GSLs) gangliotriaosylceramide (Gg(3)) and gangliotetraosylceramide (Gg(4)) have been implicated as receptors for type IV pili (T4P)-mediated Pseudomonas aeruginosa epithelial cell attachment. Since P. aeruginosa T4P are divided into five groups, the authors determined whether GSLs in general, and
Kamruddin Ahmed et al.
Medical microbiology and immunology, 191(1), 5-10 (2002-07-26)
Moraxella catarrhalis is an important pathogen of respiratory and middle ear infections. We previously reported that the attachment of M. catarrhalis to pharyngeal epithelial cells is mediated by ganglioside M2 (GM2). Several sets of adhesins or receptors are involved in
K Tadano-Aritomi et al.
Glycoconjugate journal, 13(2), 285-293 (1996-04-01)
A novel mono-sulfated glycosphingolipid based on the gangliotriaose core structure was isolated from rat kidney. The isolation procedure involved extraction of lipids with chloroform/methanol, mild alkaline methanolysis, column chromatographies with anion exchangers and silica beads. The structure was characterized by
Annelies Coddens et al.
PloS one, 6(9), e23309-e23309 (2011-09-29)
Enterotoxigenic F4-fimbriated Escherichia coli is associated with diarrheal disease in neonatal and postweaning pigs. The F4 fimbriae mediate attachment of the bacteria to the pig intestinal epithelium, enabling an efficient delivery of diarrhea-inducing enterotoxins to the target epithelial cells. There

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service