Skip to Content
Merck
All Photos(1)

Key Documents

G8671

Sigma-Aldrich

PDGFRβ(557-end), active, GST tagged human

PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

CD140B, JTK12, PDGFR1

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
$537.00

$537.00


Estimated to ship on19 May 2025


Request a Bulk Order

Select a Size

Change View
10 μG
$537.00

About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

$537.00


Estimated to ship on19 May 2025


Request a Bulk Order

recombinant

expressed in baculovirus infected Sf9 cells

Quality Level

product line

PRECISIO® Kinase

Assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

specific activity

16-22 nmol/min·mg

mol wt

~104 kDa

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... PDGFRB(5159)

General description

PDGFRB (platelet derived growth factor receptor β) is a type III tyrosine kinase receptor, which exists as a homodimer. This receptor interacts with multiple ligands, but its main physiological ligand is PDGF-BB homodimer.[1]

Biochem/physiol Actions

PDGFRB (platelet derived growth factor receptor β), upon interaction with PGDFB, regulates multiple processes such as, sclerotic disorders, wound healing and tumor angiogenesis. Gain-of-function mutation in this gene is linked with Penttinen syndrome, which is an autosomal-dominant disorder.[1] Release of PGDFB by endothelial cells results in the activation of this receptor, which is essential for the recruitment of PDGFRB+ vascular smooth muscle cells and pericytes during blood vessel formation. Loss-of-function mutation in this gene results in the neurodegenerative disorder primary familial brain calcification (PFBC).[2]

Physical form

Supplied in 50 mM Tris-HCl, pH 7.5, with 150 mM NaCl, 0.2 5mM DTT, 0.1 mM EGTA, 0.1 mM EDTA, 0.1 mM PMSF, and 25% glycerol.

Legal Information

PRECISIO is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lei Xu et al.
Cancer research, 65(13), 5711-5719 (2005-07-05)
Renal cell carcinoma is a highly malignant and often fatal disease of the kidney. It is difficult to treat, often because metastases are common at the time of presentation. Platelet-derived growth factor-D (PDGF-D) is a newly discovered member of the
P Soriano
Genes & development, 8(16), 1888-1896 (1994-08-15)
Platelet-derived growth factor, a major mitogen and chemoattractant for a number of cell types, is implicated in the processes of wound healing, tumorigenesis, and differentiation and is recognized by two receptors, alpha and beta. To begin understanding the role of
Michael Vanlandewijck et al.
PloS one, 10(11), e0143407-e0143407 (2015-11-26)
Primary Familial Brain Calcification (PFBC), a neurodegenerative disease characterized by progressive pericapillary calcifications, has recently been linked to heterozygous mutations in PDGFB and PDGFRB genes. Here, we functionally analyzed several of these mutations in vitro. All six analyzed PDGFB mutations
Jennifer J Johnston et al.
American journal of human genetics, 97(3), 465-474 (2015-08-19)
Penttinen syndrome is a distinctive disorder characterized by a prematurely aged appearance with lipoatrophy, epidermal and dermal atrophy along with hypertrophic lesions that resemble scars, thin hair, proptosis, underdeveloped cheekbones, and marked acro-osteolysis. All individuals have been simplex cases. Exome

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service