Skip to Content
Merck
All Photos(1)

Key Documents

G7754

Sigma-Aldrich

L-Glutamic acid γ-(α-naphthylamide) monohydrate

Sign Into View Organizational & Contract Pricing

Select a Size

50 MG
$114.00
1 G
$992.00

$114.00


Estimated to ship on09 May 2025


Request a Bulk Order

Select a Size

Change View
50 MG
$114.00
1 G
$992.00

About This Item

Empirical Formula (Hill Notation):
C15H16N2O3 ·H2O
CAS Number:
Molecular Weight:
290.31
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.83

$114.00


Estimated to ship on09 May 2025


Request a Bulk Order

Assay

≥98% (TLC)

Quality Level

form

powder

solubility

1 N NaOH: ethanol (1:1): 50 mg/mL, clear, colorless to orange

storage temp.

−20°C

SMILES string

O.N[C@@H](CCC(=O)Nc1cccc2ccccc12)C(O)=O

InChI

1S/C15H16N2O3.H2O/c16-12(15(19)20)8-9-14(18)17-13-7-3-5-10-4-1-2-6-11(10)13;/h1-7,12H,8-9,16H2,(H,17,18)(H,19,20);1H2/t12-;/m0./s1

InChI key

QJIBNSSNADIAKZ-YDALLXLXSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Carc. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service