Skip to Content
Merck
All Photos(1)

Key Documents

G6173

Sigma-Aldrich

GSK 4716

≥98% (HPLC)

Synonym(s):

2-[[4-(1-Methylethyl)phenyl]methylene]hydrazide 4-hydroxybenzoic acid

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$147.00
50 MG
$589.00

$147.00


Estimated to ship on01 June 2025


Request a Bulk Order

Select a Size

Change View
10 MG
$147.00
50 MG
$589.00

About This Item

Empirical Formula (Hill Notation):
C17H18N2O2
CAS Number:
Molecular Weight:
282.34
MDL number:
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.77

$147.00


Estimated to ship on01 June 2025


Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to off-white

solubility

DMSO: >10 mg/mL

originator

GlaxoSmithKline

storage temp.

2-8°C

SMILES string

CC(C)c1ccc(cc1)\C=N\NC(=O)c2ccc(O)cc2

InChI

1S/C17H18N2O2/c1-12(2)14-5-3-13(4-6-14)11-18-19-17(21)15-7-9-16(20)10-8-15/h3-12,20H,1-2H3,(H,19,21)/b18-11+

InChI key

IKPPIUNQWSRCOZ-WOJGMQOQSA-N

Biochem/physiol Actions

GSK 4716 is an Estrogen-related receptors ERRβ and ERRγ agonist with minimal activity for ERRα, ERα and ERβ

Features and Benefits

This compound is featured on the Nuclear Receptors (Steroids) page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.
This compound was developed by GlaxoSmithKline. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

We offers many products related to Nuclear Receptors (Steroids) for your research needs.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service