Skip to Content
Merck
All Photos(1)

Key Documents

G6132

Sigma-Aldrich

Glycodeoxycholic acid monohydrate

≥97% (TLC)

Synonym(s):

3α,12α-Dihydroxy-5β-cholanoic acid N-(carboxymethyl)amide, Glycodesoxycholic acid, N-(3α,12α-Dihydroxy-24-oxocholan-24-yl)glycine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C26H43NO5
CAS Number:
Molecular Weight:
467.64
Beilstein:
2954947
MDL number:
UNSPSC Code:
12161900
PubChem Substance ID:

description

anionic

Assay

≥97% (TLC)

SMILES string

[H]O[H].[H][C@]12CC[C@@]3([H])[C@]4([H])CC[C@H]([C@H](C)CCC(=O)NCC(O)=O)[C@@]4(C)[C@@H](O)C[C@]3([H])[C@@]1(C)CC[C@@H](O)C2

InChI

1S/C26H43NO5.H2O/c1-15(4-9-23(30)27-14-24(31)32)19-7-8-20-18-6-5-16-12-17(28)10-11-25(16,2)21(18)13-22(29)26(19,20)3;/h15-22,28-29H,4-14H2,1-3H3,(H,27,30)(H,31,32);1H2/t15-,16-,17-,18+,19-,20+,21+,22+,25+,26-;/m1./s1

InChI key

DLOTUJUSJVIXDW-YEUHZSMFSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Induces apoptosis in hepatocytes; may induce DNA cleavage.

replaced by

Product No.
Description
Pricing

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Sandeep Khurana et al.
PloS one, 7(2), e32006-e32006 (2012-02-24)
Bile acids (BAs) regulate cardiovascular function via diverse mechanisms. Although in both health and disease serum glycine-conjugated BAs are more abundant than taurine-conjugated BAs, their effects on myogenic tone (MT), a key determinant of systemic vascular resistance (SVR), have not
Yong Zhou et al.
American journal of physiology. Gastrointestinal and liver physiology, 298(5), G722-G731 (2010-03-06)
The injurious effect of nonsteroidal anti-inflammatory drugs (NSAIDs) in the small intestine was not appreciated until the widespread use of capsule endoscopy. Animal studies found that NSAID-induced small intestinal injury depends on the ability of these drugs to be secreted
Lutz Schneider et al.
Pancreas, 38(6), 676-680 (2009-07-25)
Calcitonin gene-related peptide (CGRP) is released from perivascular pancreatic nerves. It effects vasomotion and cytokine liberation in inflammatory processes, including acute pancreatitis (AP). Calcitonin gene-related peptide liberation is stimulated by capsaicin, a substance of red hot chili peppers. Aim of
Jesper Østergaard et al.
Journal of separation science, 32(10), 1712-1721 (2009-05-28)
The practical and theoretical problems associated with interpretation of binding isotherms obtained by the investigation of weak analyte-ligand interactions using mobility shift affinity CE were investigated with special emphasis on various correction methods to compensate for media effects due to
Kai A Bachmann et al.
Critical care (London, England), 17(6), R281-R281 (2013-12-10)
Severe acute pancreatitis is still a potentially life threatening disease with high mortality. The aim of this study was to evaluate the therapeutic effect of thoracic epidural anaesthesia (TEA) on survival, microcirculation, tissue oxygenation and histopathologic damage in an experimental

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service