Skip to Content
Merck
All Photos(1)

Key Documents

G6014

Sigma-Aldrich

rac-Glycerol 1-phosphate disodium salt hydrate

≥75% (enzymatic)

Synonym(s):

DL-α-Glycerophosphate disodium salt hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3H7Na2O6P · xH2O
CAS Number:
Molecular Weight:
216.04 (anhydrous basis)
Beilstein:
5177716
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:

Assay

≥75% (enzymatic)

impurities

~6 mol/mol water

storage temp.

2-8°C

SMILES string

O.[Na+].[Na+].OCC(O)COP([O-])([O-])=O

InChI

1S/C3H9O6P.2Na.H2O/c4-1-3(5)2-9-10(6,7)8;;;/h3-5H,1-2H2,(H2,6,7,8);;;1H2/q;2*+1;/p-2

InChI key

OFNNKPAERNWEDD-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application

rac-Glycerol 1-phosphate does not affect the growth and metabolism of E. coli at concentrations of 5-100μm.

Biochem/physiol Actions

rac-Glycerol 1-phosphate was shown to be produced in mammalian tissues by the hydrolysis of rac-glycerol 1:2-cyclic phosphate by a soluble phosphodiesterase.

Quality

The main impurity is β-glycerophosphate.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Michele Alves-Bezerra et al.
Biochimica et biophysica acta, 1821(12), 1462-1471 (2012-08-21)
Although triacylglycerol (TAG) stores play a critical role in organisms, mechanisms underlying TAG synthesis are poorly understood in invertebrates. In mammals, the synthesis of glycerolipids, including TAG, diacylglycerol (DAG) and phospholipids (PL), occurs predominantly by the glycerol-3-phosphate (G3P) pathway in
Jean-Louis Gallis et al.
Pharmacological research, 65(3), 387-395 (2012-01-10)
The effects of trans-resveratrol (RSV) combined with ethanol (EtOH) were evaluated by (31)P NMR on total ATP and sn-glycerol-3-phosphate (sn-G3P) contents measured in real time in isolated and perfused whole liver of the rat. Mitochondrial ATP turnover was assessed by
Sanjeewa Gamagedara et al.
Bioanalysis, 4(10), 1175-1183 (2012-06-02)
Urinary metabolomic profiles have recently drawn a lot of attention owing to a debate regarding their possible role as potential clinical markers for prostate cancer. In this study, levels of proline, kynurenine, uracil and glycerol-3-phosphate in 126 patients with genitourinary
Haruo Shimada et al.
Biochemistry, 50(19), 4114-4120 (2011-04-09)
The structure of membrane lipids in Archaea is different from those of Bacteria and Eucarya in many ways including the chirality of the glycerol backbone. Until now, heterochiral membranes were believed to be unstable; thus, no cellular organism could have
Ying Peng et al.
International journal of pharmaceutics, 441(1-2), 482-490 (2012-11-20)
Chitosan/glycerophosphate disodium (GP) thermosensitive hydrogels were prepared for the sustained delivery of venlafaxine hydrochloride (VH) and optimization of this formulation was mainly studied. Release mechanism was investigated by applying various mathematical models to the in vitro release profiles. Overall, drug

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service