Skip to Content
Merck
All Photos(1)

Key Documents

G5640

Sigma-Aldrich

Globulins Cohn fraction II, III from goat

Synonym(s):

Vitamin D binding protein

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$267.00

$267.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$267.00

About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.61

$267.00


Check Cart for Availability

Request a Bulk Order

biological source

goat

Quality Level

Assay

73-90% protein basis (biuret)

form

lyophilized powder

composition

Protein, 73-90% biuret

solubility

H2O: soluble 25 mg/mL

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

Vitamin D binding protein (DBP) is a polymorphic protein that specifically binds to vitamin D and its metabolites. This protein belongs to the albuminoid family and is highly conserved among vertebrates. The gene encoding DBP is mapped to human chromosome 4q11-q13. It is predominantly produced in the liver and is also expressed in the kidney, endocrine pancreatic cells, fat cells, and testis.

Application

Globulins were used in Biotin/avidin formulations.

Biochem/physiol Actions

Vitamin D binding protein (DBP) specifically binds and transports vitamin D and its metabolites with high affinity. It promotes fibrillar actin clearance by binding to actin monomers. DBP also binds to unsaturated fatty acids, megalin-cubilin receptor complex, the membrane of leukocytes, and the activation of the complement C5 system. The levels of DBP indicate the free concentration of all vitamin D metabolites. Decreased levels of serum DBP are observed in liver diseases, nephrotic syndrome, malnutrition, severe acute trauma, or disease.

Other Notes

Primarily γ-globulins

Physical form

Lyophilized powder containing NaCl

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service