All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C7H10Cl3NO4
CAS Number:
Molecular Weight:
278.52
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
Assay
>99% (TLC)
SMILES string
NC(CCC(=O)OCC(Cl)(Cl)Cl)C(O)=O
InChI
1S/C7H10Cl3NO4/c8-7(9,10)3-15-5(12)2-1-4(11)6(13)14/h4H,1-3,11H2,(H,13,14)
InChI key
ZKAVMZAGITUCAX-UHFFFAOYSA-N
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service