All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C6H12O6
CAS Number:
Molecular Weight:
180.16
Beilstein:
1724615
EC Number:
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
Recommended Products
Assay
≥99% (GC)
form
powder
mp
150-152 °C (lit.)
solubility
H2O: 2g + 15 mL, clear, colorless to very faintly yellow
suitability
suitable for component for culture media
SMILES string
OC[C@H]1O[C@H](O)[C@H](O)[C@@H](O)[C@@H]1O
InChI
1S/C6H12O6/c7-1-2-3(8)4(9)5(10)6(11)12-2/h2-11H,1H2/t2-,3-,4+,5-,6+/m1/s1
InChI key
WQZGKKKJIJFFOK-DVKNGEFBSA-N
Gene Information
human ... PYGM(5837)
Looking for similar products? Visit Product Comparison Guide
Analysis Note
Tested in M9 minimal salts medium as sole carbon source for Escherichia coli (ATCC 25922).
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Ana Paula Arruda et al.
Nature medicine, 20(12), 1427-1435 (2014-11-25)
Proper function of the endoplasmic reticulum (ER) and mitochondria is crucial for cellular homeostasis, and dysfunction at either site has been linked to pathophysiological states, including metabolic diseases. Although the ER and mitochondria play distinct cellular roles, these organelles also
Natalia Felitsyn et al.
Journal of neurochemistry, 106(5), 2068-2079 (2008-07-31)
Delta-aminolevulinic acid (delta-ALA) is a heme precursor implicated in neurological complications associated with porphyria and tyrosinemia type I. Delta-ALA is also elevated in the urine of animals and patients treated with the investigational drug dichloroacetate (DCA). We postulated that delta-ALA
Evelyn Fessler et al.
Molecular cancer, 14, 157-157 (2015-08-19)
Glioblastoma multiforme (GBM) is a rapidly growing malignant brain tumor, which has been reported to be organized in a hierarchical fashion with cancer stem cells (CSCs) at the apex. Recent studies demonstrate that this hierarchy does not follow a one-way
Rulan Bai et al.
Reproduction (Cambridge, England), 148(2), 119-127 (2014-05-08)
Following bidirectional communication, the conceptus and the uterine epithelium must establish a proper cell-cell interaction, resulting in the progression of implantation processes. To clarify the mechanism of conceptus attachment to the uterine endometrium, we studied whether vascular cell adhesion molecule
Lin Xiao et al.
Angiogenesis, 17(3), 511-518 (2013-11-22)
Long-term, in vitro propagation of tumor-specific endothelial cells (TEC) allows for functional studies and genome-wide expression profiling of clonally derived, well-characterized subpopulations. Using a genetically engineered mouse model of mammary adenocarcinoma, we have optimized an isolation procedure and defined growth
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service