Skip to Content
Merck
All Photos(1)

Key Documents

G211000

Sigma-Aldrich

DMT-2′O-TBDMS-rG(ib) Phosphoramidite

Synonym(s):

N-(2-methyl-1-oxopropyl)-5′-O-[bis(4-methoxyphenyl)phenylmethyl]-2′-O-[(1,1-dimethylethyl)dimethylsilyl]-guanosine, 3′-[2-cyanoethyl N,N-bis(1-methylethyl)phosphoramidite]

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
$11,110.00
500 G
$47,840.00

$11,110.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 G
$11,110.00
500 G
$47,840.00

About This Item

Empirical Formula (Hill Notation):
C50H68N7O9PSi
CAS Number:
Molecular Weight:
970.18
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.51

$11,110.00


Check Cart for Availability

Request a Bulk Order

biological source

non-animal source (no BSE/TSE risk)

Quality Level

product line

Proligo Reagents

Assay

≥99% (31P-NMR)
≥99.0% (reversed phase HPLC)

form

powder

technique(s)

oligo synthesis: suitable

impurities

≤0.3 wt. % water content (Karl Fischer)
≤0.5% P(III) Impurities 100-169ppm (31P-NMR)
≤0.5% single Impurity (reversed phase HPLC)
≤3 wt. % residual Solvent content

color

white to off-white

λ

conforms (UV/VIS Identity)

suitability

conforms to structure for H-NMR
conforms to structure for LC-MS

nucleoside profile

base: guanosine
base protecting group: isobutyryl
2' protecting group: TBDMS
5' protecting group: DMT
deprotection: standard

storage temp.

−20°C

SMILES string

COc1ccc(cc1)C(OC[C@H]2O[C@H]([C@H](O[Si](C)(C)C(C)(C)C)[C@@H]2OP(OCCC#N)N(C(C)C)C(C)C)n3cnc4C(=O)NC(NC(=O)C(C)C)=Nc34)(c5ccccc5)c6ccc(OC)cc6

InChI

1S/C50H68N7O9PSi/c1-32(2)45(58)54-48-53-44-41(46(59)55-48)52-31-56(44)47-43(66-68(12,13)49(7,8)9)42(65-67(63-29-17-28-51)57(33(3)4)34(5)6)40(64-47)30-62-50(35-18-15-14-16-19-35,36-20-24-38(60-10)25-21-36)37-22-26-39(61-11)27-23-37/h14-16,18-27,31-34,40,42-43,47H,17,29-30H2,1-13H3,(H2,53,54,55,58,59)/t40-,42-,43-,47-,67?/m1/s1

InChI key

PGJKESPHANLWME-FTZVBZPOSA-N

Looking for similar products? Visit Product Comparison Guide

General description

DMT-2′O-TBDMS-rG(ib) Phosphoramidite belongs to the group of 2′O-TBDMS RNA Phosphoramidites.

Application

RNA interference (RNAi) has become a popular tool for the sequence-specific inhibition of gene expression and can be used in target validation and other drug development techniques. The most convenient method to provide sequence-specific RNA oligonucleotides is a chemical synthesis on a solid support with RNA phosphoramidites and RNA CPG, analogous to DNA synthesis.

Features and Benefits

  • Industry standard 2′O-TBDMS protective group
  • Consistent lot-to-lot purity and performance
  • Compatible with deprotection methods based on methylamine or AMA
  • Standard RNA phosphoramidites provide excellent coupling results when used with ETT or BTT as an activator; best results are obtained with Activator 42
  • Capping with standard acetic anhydride capping reagent rather than with Fast Deprotection Cap A
  • Manufactured under a certified ISO 9001 quality system

Other Notes

The unique properties of RNA have stimulated the development of a variety of applications in diagnostics and therapeutics, as well as in basic molecular biology research where RNA can be used as:
  • Catalytic agents (ribozymes)
  • Affinity ligands (aptamers)
  • Agents to induce gene silencing (RNA interference)

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service