Skip to Content
Merck
All Photos(1)

Key Documents

G1921

Sigma-Aldrich

Glypican 3 human

recombinant, expressed in NSO cells

Sign Into View Organizational & Contract Pricing

Select a Size

50 μG
$1,070.00

$1,070.00


Estimated to ship on28 May 2025


Request a Bulk Order

Select a Size

Change View
50 μG
$1,070.00

About This Item

UNSPSC Code:
12352202
NACRES:
NA.26

$1,070.00


Estimated to ship on28 May 2025


Request a Bulk Order

biological source

human

Quality Level

recombinant

expressed in NSO cells

Assay

≥97% (SDS-PAGE)

form

lyophilized powder

UniProt accession no.

storage temp.

−20°C

Gene Information

human ... GPC3(2719)

General description

Glypican-3, a cell surface protein, is highly expressed in hepatocellular carcinoma (HCC) and few other human cancers including melanoma.[1] It is a heparan sulfate proteoglycan and is present on the cell surface. Under non-reducing conditions in SDS-PAGE, the glycanated glypican 3 appears as a smear with an apparent molecular mass of 60 - 100 kDa[2]

Biochem/physiol Actions

Sulfate proteoglycan attached to the cell surface by a glycosylphosphatidylinositol (GPI) anchor. Regulates cell survival and inhibits cell proliferation during normal development. Potential use as marker for early disease detection.

Physical form

Lyophilized from a 0.2 μm filtered solution in PBS containing 50 μg of bovine serum albumin per 1 μg of cytokine.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shiro Suzuki et al.
Human vaccines & immunotherapeutics, 10(2), 338-343 (2013-11-21)
Carcinoembryonic antigen glypican-3 (GPC3) is expressed by>40% of ovarian clear cell carcinoma (CCC) and is a promising immunotherapeutic target. We previously reported the safety of and immunological and clinical responses to a GPC3-derived peptide vaccine in a phase I clinical
Restriction enzyme cleavage of UV-irradiated DNA: a comparison of far-UV and mid-UV wavelengths.
S P Moore et al.
Photochemistry and photobiology, 45(2), 253-263 (1987-02-01)
Yu-Liang Wang et al.
World journal of gastroenterology, 18(19), 2408-2414 (2012-06-02)
To investigate the diagnostic value of glypican-3 (GPC3) and its relationship with hepatocellular carcinoma (HCC) recurrence after liver transplantation. HCC tissue samples (n = 31) obtained from patients who had undergone liver transplantation were analyzed. GPC3 mRNA and protein expression

Articles

Glycosaminoglycans are large linear polysaccharides constructed of repeating disaccharide units.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service