Skip to Content
Merck
All Photos(1)

Key Documents

G1887

Sigma-Aldrich

Gly-His-Lys acetate salt

≥97% (TLC)

Synonym(s):

Liver cell growth factor

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$317.00
50 MG
$1,170.00

$317.00


Available to ship on09 April 2025Details


Request a Bulk Order

Select a Size

Change View
10 MG
$317.00
50 MG
$1,170.00

About This Item

Linear Formula:
C14H24N6O4C2H4O2
CAS Number:
Molecular Weight:
400.43
MDL number:
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.32

$317.00


Available to ship on09 April 2025Details


Request a Bulk Order

Quality Level

Assay

≥97% (TLC)

composition

Peptide content, ~70%

storage temp.

−20°C

SMILES string

CC(O)=O.NCCCC[C@H](NC(=O)[C@H](Cc1c[nH]cn1)NC(=O)CN)C(O)=O

InChI

1S/C14H24N6O4.C2H4O2/c15-4-2-1-3-10(14(23)24)20-13(22)11(19-12(21)6-16)5-9-7-17-8-18-9;1-2(3)4/h7-8,10-11H,1-6,15-16H2,(H,17,18)(H,19,21)(H,20,22)(H,23,24);1H3,(H,3,4)/t10-,11-;/m0./s1

InChI key

MGNUTAFMLGJBGV-ACMTZBLWSA-N

Looking for similar products? Visit Product Comparison Guide

Amino Acid Sequence

Gly-His-Lys

General description

Gly-His-Lys is a tripeptide present at ∼200ng/ml in association with α-globulin and albumin, in human blood plasma.[1]

Application

Gly-His-Lys (GHK) has been used for the preparation of post-operative intra-articular injection of GHK-Cu (copper).[2]
Gly-His-Lys acetate salt has been used:
  • individually or as a complex with copper to test its effect on cytokines production in the human normal fibroblasts cell lines[3]
  • as a model peptide in N,N-dimethyl-N-methacryloxyethyl-N-(3-sulfopropyl)ammonium betaine based capillary electrophoresis separation studies[4]
  • as a component of Coon′s modified Ham′s F-12 medium for culturing Fischer rat thyroid follicular cell line, FRTL-5[5]

Biochem/physiol Actions

Gly-His-Lys (GHK) acetate salt displays affinity towards copper(II) ions. This property enables GHK to modulate the copper intake into cells.[3] GHK is also involved in quenching toxic by-products generated by lipid peroxidation.[3]

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

L Pickart et al.
Journal of chromatography, 175(1), 65-73 (1979-07-11)
Isolation and purification of growth-modulating peptides from biological sources is often accompanied by excessive losses of bioactive material. During the isolation of a growth-modulating tripeptide glycylhistidyllysine (GHL) from human plasma, copper and iron were found to co-isolate with the peptide.
Jingyuan Xiong et al.
Drug and chemical toxicology, 41(4), 501-508 (2017-11-17)
Thyroid hormones are involved in many important physiological activities including regulation of energy metabolism, development of nervous system, maintenance of cerebral functions, and so on. Endocrine-disrupting chemicals (EDCs) that interfere with thyroid functions raise serious concerns due to their frequent
Wen Jiang et al.
Analytical chemistry, 75(11), 2768-2774 (2003-09-02)
A novel capillary with covalently bonded zwitterionic surface modification was prepared by photograft polymerization of the zwitterionic monomer N,N-dimethyl-N-methacryloxyethyl-N-(3-sulfopropyl)ammonium betaine, onto the inner surface of a UV-transparent fused-silica capillary. Although the zwitterionic moieties in the resulting polymeric "tentacles" comprise both
Growth-modulating human plasma tripeptide: relationship between molecular structure and DNA synthesis in hepatoma cells.
L Pickart et al.
FEBS letters, 104(1), 119-122 (1979-08-01)
Sai-Chuen Fu et al.
Journal of orthopaedic research : official publication of the Orthopaedic Research Society, 33(7), 1024-1033 (2015-03-04)
After anterior cruciate ligament reconstruction (ACLR), the biological healing of the graft is a rate-limiting step which can contribute to graft failure. The tripeptide-copper complex glycyl-L-histidyl-L-lysine-Cu(II) (GHK-Cu) is a well-known activator of tissue remodeling. We investigated whether GHK-Cu can improve

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service