Skip to Content
Merck
All Photos(1)

Key Documents

G1296

Sigma-Aldrich

Glycitin

≥98% (HPLC)

Synonym(s):

4′,7-Dihydroxy 6-methoxyisoflavone 7-O-glucoside, 4H-1-Benzopyran-4-one, 7-(β-D-glucopyranosyloxy)-3-(4-hydroxyphenyl)-6-methoxy-, Glycitein 7-O-β-glucoside

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$224.00
25 MG
$886.00

$224.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$224.00
25 MG
$886.00

About This Item

Empirical Formula (Hill Notation):
C22H22O10
CAS Number:
Molecular Weight:
446.40
MDL number:
UNSPSC Code:
12352201
NACRES:
NA.25

$224.00


Check Cart for Availability

Request a Bulk Order

biological source

plant (Pueraria thunbergianaI)

Quality Level

Assay

≥98% (HPLC)

form

powder

technique(s)

HPLC: suitable

color

white to faint beige

mp

203 - 204  °C ((397 - 399 °F ))

solubility

10 mg, clear, colorless to faintly yellow

storage temp.

room temp

SMILES string

O=C1C2=CC(OC)=C(O[C@H]3[C@H](O)[C@@H](O)[C@H](O)[C@@H](CO)O3)C=C2OC=C1C4=CC=C(O)C=C4

InChI

1S/C22H22O10/c1-29-15-6-12-14(30-9-13(18(12)25)10-2-4-11(24)5-3-10)7-16(15)31-22-21(28)20(27)19(26)17(8-23)32-22/h2-7,9,17,19-24,26-28H,8H2,1H3/t17-,19-,20+,21-,22-/m1/s1

InChI key

OZBAVEKZGSOMOJ-MIUGBVLSSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Glycitin is an isoflavonoid recently found in flowers of Pueraria thunbergianaI. [1] Glycitin can be modified into the metabolite glycitein which has anti-inflammatory properties. [2]

Application


  • The antioxidant Glycitin protects against intervertebral disc degeneration through antagonizing inflammation and oxidative stress in nucleus pulposus cells.: This study highlights Glycitin′s therapeutic potential in mitigating intervertebral disc degeneration by counteracting inflammatory and oxidative processes, demonstrating its viability as a bioactive compound for degenerative diseases (Zhao W et al., 2023).

  • Spectrum-effect relationship study to reveal the pharmacodynamic substances in Flos Puerariae-Semen Hoveniae medicine pair for the treatment of alcohol-induced liver damage.: Analyzes the pharmacological effects of herbal components, including Glycitin, targeting liver damage recovery, illustrating the compound′s therapeutic relevance in traditional and modern medical applications (Zhang H et al., 2023).


Biochem/physiol Actions

Glycitin is a soy isoflavone in the glucoside form. A rat model study demonstrated glycitin to be effective in preventing bone loss and reversing the unfavorable changes of lipid metabolism in this model.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Other Notes

To gain a comprehensive understanding of our extensive range of Monosaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kouya Yamaki et al.
Planta medica, 68(2), 97-100 (2002-02-23)
Previously, we reported that the isoflavones tectorigenin and tectoridin, a glycosylated tectorigenin, isolated from the rhizomes of Belamcanda chinensis have an activity to inhibit prostaglandin (PG) E2 production in 12-O-tetradecanoylphorbol 13-acetate (TPA)-stimulated rat peritoneal macrophages. The inhibitory effect of tectorigenin
Isoflavone glycosides from the flowers of Pueraria thunbergiana.
Park HJ
Phytochemistry, 51, 147-151 (1999)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service