Skip to Content
Merck
All Photos(1)

Key Documents

G1260

Sigma-Aldrich

Gastrin II Sulfated

≥95% (HPLC), suitable for ligand binding assays

Sign Into View Organizational & Contract Pricing

Select a Size

0.1 MG
$433.00

$433.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
0.1 MG
$433.00

About This Item

Empirical Formula (Hill Notation):
C97H124N20O34S2
CAS Number:
Molecular Weight:
2178.27
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.26

$433.00


Check Cart for Availability

Request a Bulk Order

Product Name

Gastrin II Sulfated, ≥95% (HPLC)

Quality Level

Assay

≥95% (HPLC)

form

powder

technique(s)

ligand binding assay: suitable

color

white

UniProt accession no.

storage temp.

−20°C

SMILES string

CSCCC(NC(=O)C(Cc1c[nH]c2ccccc12)NC(=O)CNC(=O)C(Cc3ccc(OS(O)(=O)=O)cc3)NC(=O)C(C)NC(=O)C(CCC(O)=O)NC(=O)C(CCC(O)=O)NC(=O)C(CCC(O)=O)NC(=O)C(CCC(O)=O)NC(=O)C(CCC(O)=O)NC(=O)C(CC(C)C)NC(=O)C(Cc4c[nH]c5ccccc45)NC(=O)C6CCCN6C(=O)CNC(=O)C7CCC(=O)N7)C(=O)NC(CC(O)=O)C(=O)NC(Cc8ccccc8)C(N)=O

InChI

1S/C97H124N20O34S2/c1-49(2)39-68(114-95(145)71(43-54-46-100-59-18-11-9-16-57(54)59)116-97(147)73-19-12-37-117(73)76(120)48-102-85(135)60-24-30-74(118)104-60)93(143)110-65(29-35-81(129)130)91(141)109-64(28-34-80(127)128)90(140)108-63(27-33-79(125)126)89(139)107-62(26-32-78(123)124)88(138)106-61(25-31-77(121)122)87(137)103-50(3)84(134)113-69(41-52-20-22-55(23-21-52)151-153(148,149)150)86(136)101-47-75(119)105-70(42-53-45-99-58-17-10-8-15-56(53)58)94(144)111-66(36-38-152-4)92(142)115-72(44-82(131)132)96(146)112-67(83(98)133)40-51-13-6-5-7-14-51/h5-11,13-18,20-23,45-46,49-50,60-73,99-100H,12,19,24-44,47-48H2,1-4H3,(H2,98,133)(H,101,136)(H,102,135)(H,103,137)(H,104,118)(H,105,119)(H,106,138)(H,107,139)(H,108,140)(H,109,141)(H,110,143)(H,111,144)(H,112,146)(H,113,134)(H,114,145)(H,115,142)(H,116,147)(H,121,122)(H,123,124)(H,125,126)(H,127,128)(H,129,130)(H,131,132)(H,148,149,150)

InChI key

WSLIFCVCCVWYQQ-UHFFFAOYSA-N

Gene Information

human ... GAST(2520)

Amino Acid Sequence

pGlu-Gly-Pro-Trp-Leu-Glu-Glu-Glu-Glu-Glu-Ala-Tyr(SO3H)-Gly-Trp-Met-Asp-Phe-NH2

Biochem/physiol Actions

Gastrin II and Gastrin I are peptide hormones involved in gastrointestinal processes such as the stimulation of gastric (HCl) release.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

I Gede Arinton
Acta medica Indonesiana, 42(3), 142-146 (2010-08-21)
to find out biomarker as diagnostic tool of H. pylori chronic gastritis. the design of present study was a diagnostic test and there were 104 subjects with H. pylori chronic gastritis who fulfilled the inclusion and exclusion criteria. The diagnosis
T Sethi et al.
Cancer research, 52(21), 6031-6035 (1992-11-01)
Gastrin has been postulated to be a physiological growth factor, but compelling in vitro evidence of this has been difficult to obtain. In the present study we investigated whether small cell lung carcinoma cell lines could provide a useful model
A Botella et al.
Gastroenterology, 102(3), 779-786 (1992-03-01)
The aim of this study was to determine the cholecystokinin (CCK) receptor subtype involved in the direct myogenic effect of CCK on pig ileum. Smooth muscle cells were dispersed from pig ileum circular muscle layer and incubated in the presence
K Kitagawa et al.
Chemical & pharmaceutical bulletin, 49(8), 958-963 (2001-08-23)
Application of the fluoren-9-ylmethoxycarbonyl (Fmoc)-based solid-phase segment condensation approach to the preparation of sulfated peptides was investigated through the synthesis of human big gastrin-II, a 34-residue sulfated tyrosine [Tyr(SO3H)]-containing peptide. Highly acid-sensitive 2-chlorotrityl resin (Clt resin) was exclusively employed as
Saroj Kumari et al.
Chembiochem : a European journal of chemical biology, 13(2), 282-292 (2011-12-14)
Fluorescent labeling has enabled a better understanding of the relationships between receptor location, function, and life cycle. Each of these perspectives contributes new insights into drug action, particularly for G protein-coupled receptors (GPCRs). The aim of this study was to

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service