G0773
Guanosine 5′-triphosphate γ-(1-[2-nitrophenyl]ethyl) ester sodium salt
~95%
Synonym(s):
Caged GTP
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352204
Assay
~95%
solubility
DMSO: soluble
storage temp.
−20°C
Application
Used to generate GTP in situ by flash photolysis.
Caution
Protect from light and moisture.
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
J A McCray et al.
Proceedings of the National Academy of Sciences of the United States of America, 77(12), 7237-7241 (1980-12-01)
2-Nitrobenzyl derivatives have been used for several years as photolabile protecting groups in synthetic organic chemistry. Recently, P3-1-(2-nitro phenylethyladenosine 5'-triphosphate "caged ATP" was synthesized and its photolysis was shown to generate ATP in situ. This and related reactions have great
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service