Skip to Content
Merck
All Photos(1)

Documents

G007

Sigma-Aldrich

Guvacine hydrochloride

≥97% (NMR), solid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H9NO2 · HCl
CAS Number:
Molecular Weight:
163.60
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

≥97% (NMR)

form

solid

color

white

solubility

H2O: soluble
ethanol: slightly soluble

SMILES string

Cl.OC(=O)C1=CCCNC1

InChI

1S/C6H9NO2.ClH/c8-6(9)5-2-1-3-7-4-5;/h2,7H,1,3-4H2,(H,8,9);1H

InChI key

FGNUNVVTHHKDAM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

GABA uptake inhibitor.

Features and Benefits

This compound is featured on the GABA Transporters page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Preparation Note

Solutions may be stored for several days at 4 °C.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Audrey Bagnaud-Baule et al.
PloS one, 6(3), e17818-e17818 (2011-03-18)
Each year, during winter months, human Metapneumovirus (hMPV) is associated with epidemics of bronchiolitis resulting in the hospitalization of many infants. Bronchiolitis is an acute illness of the lower respiratory tract with a consequent inflammation of the bronchioles. The rapid
Xinran Xiang et al.
PloS one, 6(1), e14640-e14640 (2011-02-10)
Tumor microenvironments present significant barriers to penetration by antibodies and immunoconjugates. Tumor microenvironments, however, are difficult to study in vitro. Cells cultured as monolayers exhibit less resistance to therapy than those grown in vivo and an alternative research model more
Ximena Cortes-Bratti et al.
PloS one, 6(2), e16960-e16960 (2011-03-03)
Thioredoxin 80 (Trx80) is an 80 amino acid natural cleavage product of Trx, produced primarily by monocytes. Trx80 induces differentiation of human monocytes into a novel cell type, named Trx80-activated-monocytes (TAMs). In this investigation we present evidence for a role
Betel nut constituents as inhibitors of gamma-aminobutyric acid uptake.
G A Johnston et al.
Nature, 258(5536), 627-628 (1975-12-18)
C Rovira et al.
Neuroscience, 12(2), 543-555 (1984-06-01)
In rats under urethane anaesthesia gamma-aminobutyric acid agonists and uptake blockers were microiontophoretically applied in the pyramidal layer of CA1 and in the apical dendrites using a twin set of multibarrelled micropipettes. Thus, the somatic and dendritic field potentials elicited

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service