Skip to Content
Merck
All Photos(1)

Key Documents

FG1383

SAFC

RPMI-1640 Medium

HEPES modification, with stable glutamine, liquid, sterile-filtered, suitable for cell culture

Pharma Manufacturing

Synonym(s):

Roswell Park Memorial Institute 1640 medium

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
$24.80

$24.80


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 ML
$24.80

About This Item

UNSPSC Code:
12352207
NACRES:
NA.71

$24.80


Check Cart for Availability

Request a Bulk Order

Product Name

RPMI-1640 Medium, HEPES Modification, With 25 mM HEPES and stable glutamine, liquid, sterile-filtered, suitable for cell culture

Quality Level

sterility

sterile-filtered
sterile

form

liquid

technique(s)

cell culture | mammalian: suitable

components

stable glutamine: yes
HEPES: 25 mM
phenol red: yes
NaHCO3: yes

shipped in

ambient

storage temp.

2-8°C

Application

RPMI 1640 Medium was developed at Roswell Park Memorial Institute in 1966 by Moore and his co-workers. A modification of McCoy′s 5A Medium, it was formulated to support lymphoblastoid cells in suspension culture, but it has since been shown to support a wide variety of cells that are anchorage dependent. Originally intended to be used with a serum supplement, RPMI 1640 has been shown to support several cell lines in the absence of serum. It has also been widely used in fusion protocols and in the growth of hybrid cells.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service