Skip to Content
Merck
All Photos(1)

Key Documents

F7626

Sigma-Aldrich

6-Fluoro-DL-tryptophan

crystalline

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H11FN2O2
CAS Number:
Molecular Weight:
222.22
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

form

crystalline

mp

280-285 °C (dec.) (lit.)

storage temp.

−20°C

SMILES string

NC(Cc1c[nH]c2cc(F)ccc12)C(O)=O

InChI

1S/C11H11FN2O2/c12-7-1-2-8-6(3-9(13)11(15)16)5-14-10(8)4-7/h1-2,4-5,9,14H,3,13H2,(H,15,16)

InChI key

YMEXGEAJNZRQEH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

6-Fluoro-DL-tryptophan (6-F-TRP), a serotonin (5-HT) synthesis inhibitor, is metabolized in the brain and may be useful for tracing neuronal serotoninergic pools. 6-F-TRP is used as a competitive inhibitor of tryptophan binding to albumin and passage through the blood brain barrier (BBB).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

E Chanut et al.
Journal of neural transmission. General section, 96(2), 105-112 (1994-01-01)
6-Fluoro-serotonin (6F-5-HT) was previously identified in the rat brain after peripheral administration of 6-fluoro-DL-tryptophan, a serotonin (5-HT) synthesis inhibitor. These present studies, performed with rat brain synaptosomes show that: i-neuronal 6F-5-HT uptake partly involved the 5-HT transporter since it was
S Miwa et al.
Journal of neurochemistry, 48(5), 1577-1580 (1987-05-01)
After intraperitoneal injection of rats with 6-fluorotryptophan (6-FT), brain 5-hydroxytryptamine (5-HT) levels decreased exponentially over 1 h. Depletion was dose-dependent and maximum depletion was observed at 200 mg/kg. 6-FT (200 mg/kg) did not significantly alter the content of 5-hydroxyindoleacetic acid.
E Li et al.
The Journal of biological chemistry, 264(29), 17041-17048 (1989-10-15)
Rat cellular retinol-binding protein II (CRBP II) is a 15.6-kDa intestinal protein which binds all-trans-retinol and all-trans-retinal but not all-trans-retinoic acid. We have previously analyzed the interaction of Escherichia coli-derived rat apoCRBP II with several retinoids using fluorescence spectroscopic techniques.
F Richard et al.
Brain research, 536(1-2), 41-45 (1990-12-17)
An immunoblot procedure was developed to quantify the amount of tryptophan hydroxylase (TpOH), the rate limiting enzyme in the synthesis of serotonin, in the rat raphe dorsalis nucleus (NRD). Using this method we have studied the time course variations in
A H Baugher et al.
Biophysical journal, 75(5), 2574-2576 (1998-10-28)
Rotational-echo double-resonance (REDOR) 13C NMR spectra (with 19F dephasing) have been obtained of 6-fluorotryptophan complexed by a polymeric amphiphilic nanosphere consisting of a polystyrene core covalently attached to a poly(acrylic acid)-polyacrylamide shell. The REDOR spectra show that aromatic carbons from

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service