Skip to Content
Merck
All Photos(1)

Key Documents

F7558

Sigma-Aldrich

FSL-Tyr1

≥90% (TLC)

Synonym(s):

FSL-Tyr1(forY)-SC2-L1

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
$704.00

$704.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 MG
$704.00

About This Item

Empirical Formula (Hill Notation):
C91H140N16O33P
Molecular Weight:
2017.15
UNSPSC Code:
12352201
NACRES:
NA.26

$704.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥90% (TLC)

storage temp.

−20°C

Application

All FSL constructs disperse in biocompatible media and spontaneously and stably incorporate into cell membranes. Cells and virions modified with FSL constructs are known as kodecytes and kodevirions, respectively, and usually maintain their normal vitality and functionality. FSL-Tyr1 has been specifically designed to create radiolabeled (125I) live cells (kodecytes) but can also be used to modify other hydrophobic surfaces including fixed cells, virions, and solid phase surfaces.
All FSL constructs disperse in biocompatible media and spontaneously and stably incorporate into cell membranes. FSL-Tyr1 has been specifically designed to create radiolabeled (125I) live cells (kodecytes) but can also be used to modify other hydrophobic surfaces including fixed cells, virions, and solid phase surfaces.

Other Notes

FSL-Tyr1 is a KODE technology construct designed to label hydrophobic surfaces including living cells and virions with 125I. All FSL constructs consist of three essential designable features:
  • functional component (F)
  • spacer (S)
  • diacyl lipid (L)
FSL-Tyr1 is comprised predominantly of an N-formyl-tyrosine monomer representing F, conjugated via a spacer (S) to an an activated adipate derivative of dioleoylphosphatidylethanolamine (L).

Legal Information

KODE Constructs are supplied by KODE Biotech Materials Limited under license from KODE Biotech Limited.
KODE is a trademark of KODE Biotech Ltd.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service