Skip to Content
Merck
All Photos(1)

Key Documents

F7307

Sigma-Aldrich

Fotemustine

≥98% (HPLC)

Synonym(s):

(+-)-Diethyl (1-(3-(2-chloroethyl)-3-nitrosoureido)ethyl)phosphonate, Muphoran, Mustophorane, P-[1-[[[(2-Chloroethyl)nitrosoamino]carbonyl]amino]ethyl]-phosphonic acid diethyl ester, S 10036

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H19ClN3O5P
CAS Number:
Molecular Weight:
315.69
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

≥98% (HPLC)

form

powder

storage condition

protect from light

color

light yellow

solubility

H2O: ≥1 mg/mL

originator

Servier

storage temp.

2-8°C

SMILES string

CCOP(=O)(OCC)C(C)NC(=O)N(CCCl)N=O

InChI

1S/C9H19ClN3O5P/c1-4-17-19(16,18-5-2)8(3)11-9(14)13(12-15)7-6-10/h8H,4-7H2,1-3H3,(H,11,14)

InChI key

YAKWPXVTIGTRJH-UHFFFAOYSA-N

Biochem/physiol Actions

Fotemustine is a third generation nitrosourea, chloroethylating agent used in the treatment of glioma and malignant melanoma.

Features and Benefits

This compound is a featured product for Apoptosis research. Click here to discover more featured Apoptosis products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.
This compound was developed by Servier. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Carc. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anna Maria Di Giacomo et al.
The Lancet. Oncology, 13(9), 879-886 (2012-08-17)
Ipilimumab improves survival of patients with metastatic melanoma, many of whom develop brain metastases. Chemotherapy-induced release of tumour antigens might amplify ipilimumab's antitumour activity. We aimed to investigate the efficacy and safety of ipilimumab plus fotemustine in patients with metastatic
Xavier Durando et al.
Bulletin du cancer, 100(1), 23-28 (2012-08-15)
Brain metastases affect 37% of patients suffering from metastatic melanoma, and their prognosis remains poor, with an overall survival lower than six months. At the moment, there is no standard therapeutic strategy for management of melanoma brain metastases. In some
M Santoni et al.
Anticancer research, 32(3), 1099-1101 (2012-03-09)
Alkylating agents, such as temozolomide (TMZ) and fotemustine (FTM) are widely used in recurrent glioblastoma (GBM) regimes. Several strategies have been proposed to prevent resistance to these agents, by combining or sequencing them. We report the results of a pilot
Silvia Mangiacavalli et al.
American journal of hematology, 88(2), 102-106 (2012-12-12)
Since multiple myeloma (MM) is still not-curable, the management of relapse remains challenging. Given the known efficacy of alkylating agents in MM, we conducted a phase I/II study to test a new three drug combination in which Fotemustine (Muphoran), an
Ipilimumab with fotemustine in metastatic melanoma.
Claus Garbe
The Lancet. Oncology, 13(9), 851-852 (2012-08-17)

Articles

Cell cycle phases (G1, S, G2, M) regulate cell growth, DNA replication, and division in proliferating cells.

Apoptosis regulation involves multiple pathways and molecules for cellular homeostasis.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service