Skip to Content
Merck
All Photos(2)

Key Documents

F6807

Sigma-Aldrich

Fleroxacin

Synonym(s):

RO 23-6240, 6,8-Difluoro-1-(2-fluoroethyl)-1,4-dihydro-7-(4-methylpiperazino)-4-oxo-3-quinolinecarboxylic acid, 6,8-Difluoro-1-(2-fluoroethyl)1,4-dihydro-7-(4-methyl-1-piperazinyl)-4-oxo-3-quinolinecarboxylic acid, AM-833, Megalocin, Megalone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H18F3N3O3
CAS Number:
Molecular Weight:
369.34
MDL number:
UNSPSC Code:
51102829
PubChem Substance ID:
NACRES:
NA.85

Assay

≥98% (HPLC)

form

solid

color

white to off-white

solubility

0.1 M NaOH: 10 mg/mL

antibiotic activity spectrum

Gram-negative bacteria
Gram-positive bacteria
mycobacteria
mycoplasma

Mode of action

DNA synthesis | interferes
enzyme | inhibits

storage temp.

−20°C

SMILES string

CN1CCN(CC1)c2c(F)cc3C(=O)C(=CN(CCF)c3c2F)C(O)=O

InChI

1S/C17H18F3N3O3/c1-21-4-6-22(7-5-21)15-12(19)8-10-14(13(15)20)23(3-2-18)9-11(16(10)24)17(25)26/h8-9H,2-7H2,1H3,(H,25,26)

InChI key

XBJBPGROQZJDOJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Fleroxacin is a broad-spectrum antimicrobial fluoroquinolone. It is used in pharmacokinetic studies and is used to study the treatment of intra-abdominal abscesses and travellers′ diarrhea.

Biochem/physiol Actions

Fleroxacin is a synthetic trifluorinated quinolone with antimicrobial activity against a variety of pathogens, including mycobacteria, mycoplasmas, chlamydiae, and legionellae. It strongly inhibits the DNA-supercoiling activity of DNA gyrase and DNA topoisomerase 2, which results in cell death.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Pefanis et al.
Antimicrobial agents and chemotherapy, 38(2), 252-255 (1994-02-01)
To assess the potential efficacy of fleroxacin in combination with clindamycin or metronidazole in mixed aerobic and anaerobic infections, we used a rat model of intra-abdominal abscesses in which the inoculum consisted of pooled rat feces mixed with BaSO4. Two
R A Blouin et al.
Antimicrobial agents and chemotherapy, 36(3), 632-638 (1992-03-01)
In this open-label study, the disposition of fleroxacin in liver disease in 12 healthy male volunteers, 6 male cirrhotics without ascites (group A), and 6 male cirrhotics with ascites (group B) was evaluated. Fleroxacin (400 mg) was administered orally and
J C Akaniro et al.
Antimicrobial agents and chemotherapy, 34(10), 1880-1884 (1990-10-01)
We evaluated fleroxacin, a newer fluoroquinolone, against isolates from sputum from patients with cystic fibrosis. These isolates included rough and mucoid Pseudomonas aeruginosa, Pseudomonas cepacia, Staphylococcus aureus, Haemophilus influenzae, and Escherichia coli. Selected isolates were tested by the broth microdilution
R Steffen et al.
The Journal of antimicrobial chemotherapy, 31(5), 767-776 (1993-05-01)
A double-blind, randomized, placebo-controlled trial was conducted to evaluate the efficacy and safety of fleroxacin for one or two days as treatment for patients with travellers' diarrhoea. A total of 195 patients who were suffering with acute diarrhoea of less
Xiaozhang Zhu et al.
Journal of the American Chemical Society, 133(41), 16342-16345 (2011-09-22)
A heptacyclic carbocycle possessing three p-quinodimethane units conjugated in one plane has been synthesized and shown to exhibit distinct biradical characteristics. The molecule has a HOMO/LUMO band gap of ca. 1 eV and a S(0)-T(1) energy gap of 2.12 kcal/mol

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service