Skip to Content
Merck
All Photos(1)

Key Documents

F3554

Sigma-Aldrich

Anti-Frizzled-10 antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-FZD10

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203

biological source

rabbit

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 3 μg/mL using human kidney, cortex

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... FZD10(11211)
mouse ... Fzd10(93897)

Immunogen

synthetic peptide corresponding to the N-terminal extracellular domain of human frizzled-10, conjugated to KLH. The immunizing peptide has 100% homology with the mouse gene.

Application

Anti-Frizzled-10 antibody is suitable for immunohistochemistry study (formalin-fixed, paraffin-embedded sections) at a concentration of 3μg/mL using human kidney, cortex.

Biochem/physiol Actions

Frizzled homolog protein (FZD) is a seven-pass transmembrane type receptor expressed in various normal tissues such as skeletal muscle, kidney, pancreas, cerebellum and cerebral cortex. In humans, ten members (FZD1-FZD10) of Frizzled homolog protein have been identified. FZD10, a cell-surface receptor molecules in the Wnt pathway, is expressed during development in embryonic limbs, neural tube, and central nervous system and mediates Wnt signaling. It was reported that FZD10 plays a critical role in the metastasis of synovial sarcoma. Mechanistically, it facilitates the activation of Dvl-Rac1-JNK axis of planar cell polarity (PCP) signaling, with no effect on nuclear β-catenin accumulation and Tcf/Lef reporter activity.

Physical form

Solution in phosphate buffered saline, pH 7.7, containing 0.01% sodium azide.

Not finding the right product?  

Try our Product Selector Tool.


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Satoshi Nagayama et al.
Oncogene, 24(41), 6201-6212 (2005-07-12)
Genome-wide expression profiling revealed overexpression of the gene encoding frizzled homologue 10 (FZD 10), a cell-surface receptor for molecules in the Wnt pathway, as a potential contributor to synovial sarcomas (SS). Northern blotting and immunohistochemical staining confirmed that expression levels
Yingqun Wang
Molecular cancer therapeutics, 8(8), 2103-2109 (2009-08-13)
The evolutionarily conserved and developmentally important Wnt signaling pathway has traditionally been regarded as a critical player in tumorigenesis through the canonical Wnt/beta-catenin cascade. Nevertheless, accumulating evidence based on recent research has revealed the previously unacknowledged role of noncanonical Wnt/planar
Edlaine Linares et al.
Chemical research in toxicology, 37(7), 1129-1138 (2024-06-25)
Peroxymonocarbonate (HCO4-/HOOCO2-) is produced by the reversible reaction of CO2/HCO3- with H2O2 (K = 0.33 M-1, pH 7.0). Although produced in low yields at physiological pHs and H2O2 and CO2/HCO3- concentrations, HCO4- oxidizes most nucleophiles with rate constants 10 to
Koji Ueno et al.
International journal of cancer, 132(8), 1731-1740 (2012-07-27)
Wnt signaling pathways play important roles in tumorigenesis and are initiated by binding of Wnt to various receptors including frizzleds (FZDs). FZDs are one of several families of receptors comprised of FZD/LRP/ROR2/RYK in the Wnt signaling pathway. Expression of some
H Kirikoshi et al.
International journal of oncology, 19(4), 767-771 (2001-09-20)
Frizzled (FZD) genes encode seven-transmembrane type WNT receptors, which are implicated in carcinogenesis and embryogenesis. We have previously cloned and characterized FZD1, FZD2, FZD3, FZD4, FZD5, FZD6, FZD7, FZD8, and FZD10. Here, we investigated expression profiles of all members of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service