Skip to Content
Merck
All Photos(2)

Key Documents

F1428

Sigma-Aldrich

Fenvalerate

≥97%

Synonym(s):

α-Cyano-3-phenoxybenzyl α-(4-chlorophenyl)­iso­valerate

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
$241.00

$241.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 MG
$241.00

About This Item

Empirical Formula (Hill Notation):
C25H22ClNO3
CAS Number:
Molecular Weight:
419.90
Beilstein:
2025982
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

$241.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥97%

storage temp.

−20°C

SMILES string

CC(C)C(C(=O)OC(C#N)c1cccc(Oc2ccccc2)c1)c3ccc(Cl)cc3

InChI

1S/C25H22ClNO3/c1-17(2)24(18-11-13-20(26)14-12-18)25(28)30-23(16-27)19-7-6-10-22(15-19)29-21-8-4-3-5-9-21/h3-15,17,23-24H,1-2H3

InChI key

NYPJDWWKZLNGGM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Fenvalerate an ester molecule, exists in four stereoisomers with two chiral centers.[1]

Application

Fenvalerate has been used to study the effect of Fen exposure on reproductive functions.[2]

Biochem/physiol Actions

Fenvalerate has good insecticidal potential and is less toxic to animals.[3] It might cause endocrine disruption and reproductive dysfunction in humans.[4] Metabolism of fenvalerate includes oxidation, ester cleavage and conjugation reaction. It is stable at pH 5 and 7, while it gets hydrolyzed at pH 9.[1] In rat models it is found to hinder the action of mitochondrial enzymes.[3]
Fenvalerate is a type II semi-synthetic pyrethrin, which acts as a potent inhibitor of calcineurin (protein phosphatase 2B). This inhibitory action results in cellular hyperexcitability by causing non-mutated calcium channels to remain open for an extended period of time allowing an abundance of Ca2+ to enter the cell.
Type II pyrethroid. Potent inhibitor of calcineurin (protein phosphatase 2B).

Preparation Note

Fenvalerate has been reported to be soluble in DMSO, ethanol, and acetone.

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Metabolic Pathways of Agrochemicals: Insecticides and fungicides, 659-659 (1998)
Paternal fenvalerate exposure influences reproductive functions in the offspring
Xia D, et al.
Reproductive Sciences, 20(11), 1308-1315 (2013)
Fenvalerate-induced oxidative damage in rat tissues and its attenuation by dietary sesame oil
Prasanthi K, et al.
Food And Chemical Toxicology, 43(2), 299-306 (2005)
Effects of fenvalerate exposure on semen quality among occupational workers
Lifeng T, et al.
Contraception, 73(1), 92-96 (2006)
Tao Tang et al.
Pest management science, 68(11), 1501-1511 (2012-06-13)
The function of cytochrome P450 proteins (P450s) in the metabolism of a variety of compounds by oxidation and reduction is well elucidated, but its interactions with other electron transfer components in the pyrethroid resistance of insect pests have been a

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service