Key Documents
EPAG7-TT20
Test Tube Rack, for Eppendorf® New Brunswick Innova® 3100 Water Bath Shakers
holds 29 x 20 mm tubes
About This Item
UNSPSC Code:
41122804
NACRES:
NB.73
Product EPAG7-TT20 is not currently sold in your country. Contact Technical Service
material
stainless steel
feature
holds 29 x 20 mm tubes
manufacturer/tradename
Eppendorf® AG7-TT20 (North America)
General description
Test Tube Racks have an adjustable angle and tube height ­support. Made from stainless-steel with inserts to help retain tubes and ­prevent them from spinning in the rack. Finger-tight knobs let you quickly adjust angle, or release the rack from the base plate, for easy transport to the bench.
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Innova is a registered trademark of Eppendorf, Inc. 
New Brunswick is a trademark of Eppendorf AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)