EPAG-2
Platform for Eppendorf® New Brunswick™ Innova® 2300/43/43R, Excella® E10/E25/E25R Shakers
for 2 L Erlenmeyer flasks, dedicated, aluminum
About This Item
material
aluminum
feature
dedicated
for 2 L Erlenmeyer flasks
manufacturer/tradename
Eppendorf® AG-2 (Global)
size
76 cm × 46 cm
capacity
(12 x 2 L Erlenmeyer flasks)
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service