EP6731010515
Eppendorf® CellXpert® C170i CO2 Incubator
Std, Right, copper, AC/DC input 120 V, US 2-pin plug
All Photos(1)
About This Item
Recommended Products
feature
Std, Right
manufacturer/tradename
6731010515
Eppendorf®
parameter
4-50 °C temp. range
capacity
(170 L)
(740 L)
color
copper
Related Categories
General description
Stay flexible for the future
Safety for your cells
Save money
- Upgrade your device later in your lab (e.g. door handle position)
- Advanced user interface for easy and complete control over parameter settings, monitoring, and documentation
Safety for your cells
- Uniform temperature with multiple sensors verified at 27 points (German Norm DIN 12880)
- Fast temperature and CO2 recovery in less than 5 minutes without overshoot
- Vibration and turbulence protection with fan-less design
- 180 ºC High Temperature Disinfection (HTD)
Save money
- Significantly reduced gas consumption with smart gas control
- Up to 25 % more usable space with fan-less design
- No recurring costs for internal replacement parts (HEPA filters, UV lamps etc.)
Legal Information
CellXpert is a registered trademark of Eppendorf AG
Eppendorf is a registered trademark of Eppendorf AG